Search
Make sure you’re operating your Thermo Scientific NanoDrop Microvolume Spectrophotometers and Fluorometers with full functionality. Based on input from customers like you, we are constantly updating and evolving NanoDrop software to deliver greater capabilities and efficiencies in the lab.
You can download and install the latest operating software by following the steps in this section. Minimum computer and operating system requirements needed to run each NanoDrop instrument are described in the installation section.
Tip: If you are using a NanoDrop Ultra or NanoDrop Eight instrument in a regulated environment, you can also purchase optional compliance software that helps you meet FDA 21 CFR Part 11 requirements for electronic recordkeeping. Visit the NanoDrop Ultra or NanoDrop Eight catalog page for more information or simply order below.
Registration is a one-time process that allows you to receive alerts when new software is available for your instrument, and helps verify your warranty status.
When you register, make sure to check the box allowing marketing emails from Thermo Fisher Scientific, as we cannot alert you to new updates without your permission.
The table lists the latest operating software versions available for your instrument. To check your current version, select System status (on your NanoDrop Ultra/UltraC, Ultra FL/UltraC FL, NanoDrop One/OneC or NanoDrop Eight instrument) or select Help > About from the menu in the software on your PC (for all other instruments).
Latest operating software for NanoDrop instruments
Registration is a one-time process that allows you to receive alerts when new software is available for your instrument, and helps verify your warranty status.
When you register, make sure to check the box allowing marketing emails from Thermo Fisher Scientific, as we cannot alert you to new updates without your permission.
Instrument |
Software module |
Version |
Release date |
Download |
|---|---|---|---|---|
NanoDrop Ultra/Ultraᶜ/Ultra FL/Ultraᶜ FL |
Local Control Software | 1.3.0 |
December 2025 | |
| PC Control Software | 1.3.0 | December 2025 | ||
NanoDrop Eight |
Operating Software |
1.8 |
December 2025 | |
NanoDrop Lite Plus |
Operating Software |
1.2.2 |
July 2025 |
|
| NanoDrop One/Oneᶜ | Local Control Software | 2.12.0 | October 2024 | |
| PC Control Software | 2.12.0 |
October 2024 | ||
NanoDrop 3300 |
Operating Software |
2.8.1 |
August 2020 |
|
NanoDrop 2000/2000c |
Operating Software |
1.6 |
September 2014 |
|
NanoDrop 8000 |
Operating Software |
2.3.3 |
April 2020 |
|
NanoDrop 1000 |
Operating Software |
3.8.1 |
June 2011 |
To download the latest version:
Once the file(s) are downloaded to your computer, come back to this page and refer to the appropriate section for installation instructions.
The NanoDrop Eight spectrophotometer is controlled from a Windows PC connected by a USB cable. The operating software is compatible with Windows 10/11 Professional and Enterprise Build 1607 or higher.
You must be an administrator on the computer to install the software. Remember to disconnect the instrument from the computer while you download and install the software on your computer.
To install a custom method to your NanoDrop Eight instrument, click the link below to download the method file to your PC.
Install software on your NanoDrop Lite Plus instrument via a USB drive:
Because the NanoDrop One/OneC spectrophotometer can be controlled from either its local touchscreen or from a connected PC, you will need to install software update files for both.
Important: To ensure data transfer and viewing, the software versions for local instrument and PC control must match. For example, if you are updating your local NanoDrop One Operating and help software to version 2.10 (March 2023), make sure to update your PC Control software to version 2.10 (March 2023) as well.
You can create a custom method to analyze your own samples in your own way or make use of several custom methods that we’ve predefined. To install a custom method to your NanoDrop One/OneC instrument, click its link below to download the method file to your PC.
Predefined NanoDrop One/OneC custom methods
NanoDrop One PC Control software gives you the flexibility to control the operation of your instrument from an Ethernet-connected PC. The software is compatible with Windows 10/11 Pro (64 bit) or higher.
The NanoDrop 3300 operating software is compatible with Windows 7 Professional (32 bit and 64 bit), Windows 8.1 (64 bit), and Windows 10/11 (64 bit).
You must be an administrator on the computer to install the software. Remember to disconnect the instrument from the computer while you download and install the software on your computer.
Prior to installing the latest software version, all previous versions of NanoDrop 3300 and associated software must be uninstalled.
The NanoDrop 2000/2000c operating software is compatible with Windows 7 Professional (32 bit and 64 bit), Windows 8.1 Professional (64 bit), and Windows 10/11 Pro (64 bit).
You must be an administrator on the computer to install the software.
The NanoDrop 8000 operating software is compatible with Windows 7 Professional (32 bit and 64 bit), Windows 8.1 (64 bit), and Windows 10/11 (64 bit).
You must be an administrator on the computer to install the software. Remember to disconnect the instrument from the computer when you download the software.
The NanoDrop 1000 operating software is compatible with Windows 7 Professional (32 bit and 64 bit), Windows Vista (32 bit), and Windows XP (32 bit).
You must be an administrator on the computer to install the software.
If you are using a NanoDrop One/OneC or NanoDrop Eight instrument in a regulated environment, you can also purchase optional compliance software that helps you meet FDA 21 CFR Part 11 requirements for electronic recordkeeping. The modules let you control user account access, apply digital signatures, and review audit lots, and can be installed on a central computer to manage settings across multiple instruments.
Order Security Suite software (for the NanoDrop One/OneC instrument) or SciVault software (for the NanoDrop Eight instrument) below, or follow the links in the table for more information.
For Research Use Only. Not for use in diagnostic procedures.