EZ-Link™ BMCC-Biotin
Thermo Scientific™

EZ-Link™ BMCC-Biotin

Thermo Scientific EZ-Link BMCC-Biotin is a maleimide-activated, sulfhydryl-reactive biotinylation reagent with an extended spacer arm that contains a stabilizing cyclohexaneRead more
Have Questions?
Catalog NumberQuantity
2190050 mg
Catalog number 21900
Price (USD)
362.00
Each
Quantity:
50 mg
Price (USD)
362.00
Each
Thermo Scientific EZ-Link BMCC-Biotin is a maleimide-activated, sulfhydryl-reactive biotinylation reagent with an extended spacer arm that contains a stabilizing cyclohexane group.

Features of EZ-Link BMCC-Biotin:

Protein labeling—biotinylate antibodies or other proteins for use in protein methods
Membrane-permeable—can be used to label inside cells (intracellular)
Thiol-reactive—reacts with sulfhydryls (-SH), such as the side-chain of cysteine (C)
Maleimide-activated—perform reactions at pH 6.5 to 7.5 in buffers such as PBS
Irreversible—forms permanent thioether bonds; spacer arm cannot be cleaved
Solubility—must be dissolved in DMSO or DMF before further dilution in aqueous buffers
Medium length—spacer arm (total length added to target) is 32.6 angstroms; contains cyclohexane ring, which stabilizes adjacent maleimide

BMCC-Biotin is a maleimido-biotin compound for labeling protein cysteines and other molecules that contain sulfhydryl groups. This reagent specifically reacts with reduced thiols (-SH) in near-neutral buffers to form permanent (irreversible) thioether bonds. The unique feature of BMCC-Biotin is its spacer arm cyclohexane ring; this has a stabilizing effect that minimizes hydrolysis and degradation of the maleimide group until it has opportunity to conjugate with target thiols.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.

Biotinylation reagents differ in reactivity, length, solubility, cell permeability and cleavability. Three types of sulfhydryl-reactive compounds are available: maleimido, iodoacetyl and pyridyldithiol. Maleimide reagents specifically react with sulfhydryl groups (-SH) in near-neutral buffers to form permanent thioether bonds.

In proteins, sulfhydryls exist where there are cysteine (C) residues. Cystine disulfide bonds must be reduced to make sulfhydryl groups available for labeling. Hinge-region disulfide bridges of antibodies can be selectively reduced to make functional half-antibodies that can be labeled.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-Permeant
Label TypeBiotin & Analogs
Product LineEZ-Link
Product TypeBMCC-Biotin
Quantity50 mg
Reactive MoietyMaleimide
Chemical ReactivityThiol
Label or DyeBiotin
SolubilityDMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
SpacerExtra-long
Unit SizeEach
Contents & Storage
Store at 4°C.

Frequently asked questions (FAQs)

What is the advantage of using EZ-Link HPDP-Biotin over maleimide- or iodoacetyl-containing biotinylation reagents?

Both maleimide- and iodoacetyl-containing biotinylation reagents react with sulfhydryl (-SH) groups to form stable thioether bonds that are not cleavable. EZ-Link HPDP-Biotin on the other hand is a pyridyldithiol-biotin compound that reacts with -SH groups in near-neutral buffers to form reversible disulfide bonds. Because the disulfide group can be cleaved using DTT or other reducing agents, HPDP-Biotin is useful for labeling and affinity-purification applications that require recovery of the original, unmodified molecule.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (1)

Citations & References
Abstract
Rif1 S-acylation mediates DNA double-strand break repair at the inner nuclear membrane.
Authors:Fontana GA, Hess D, Reinert JK, Mattarocci S, Falquet B, Klein D, Shore D, Thomä NH, Rass U
Journal:Nat Commun
PubMed ID:31182712
'Rif1 is involved in telomere homeostasis, DNA replication timing, and DNA double-strand break (DSB) repair pathway choice from yeast to human. The molecular mechanisms that enable Rif1 to fulfill its diverse roles remain to be determined. Here, we demonstrate that Rif1 is S-acylated within its conserved N-terminal domain at cysteine ... More