1-Step™ TMB-Blotting Substrate Solution
Thermo Scientific™
1-Step™ TMB-Blotting Substrate Solution
Thermo Scientific Pierce TMB-Blotting 1-Step Solution is a ready-to-use solution of the blue TMB peroxidase (HRP) substrate that is speciallyRead more
Have Questions?
Catalog NumberQuantity
34018250 mL
Catalog number 34018
Price (USD)
210.00
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
210.00
Each
Thermo Scientific Pierce TMB-Blotting 1-Step Solution is a ready-to-use solution of the blue TMB peroxidase (HRP) substrate that is specially formulated for chromogenic detection in Western blot and IHC experiments.
TMB (3,3',5,5'-tetramethylbenzidine; MW = 240.4) is most often used as a soluble substrate to detect the activity of peroxidase-conjugated antibodies in ELISA. Pierce TMB-Blotting Solution is formulated to precipitate and localize the blue reaction product onto membrane surfaces or tissue samples at sites where the peroxidase-substrate reaction occurs. TMB-Blotting Substrate produces a very large signal-to-noise ratio, although its high sensitivity requires careful washing and blocking to avoid background staining. When performing immunohistochemical techniques, aqueous counterstains and mounting medium must be used.
Features of 1-Step TMB-Blotting Solution:
HRP substrate—for sensitive detection of horseradish peroxidase activity on solid media
Chromogenic—no special equipment needed for visualization; produces dark blue bands in Western blots; is not as susceptible to fading as AEC or 4-CN substrates
Ready-to-use—no organic solvents are required to dissolve; no dilution necessary for use
Complete—does not require other buffers or addition of hydrogen peroxide
Requires:
• For best results, use TMB-Blotting Substrate with blots that have been blocked with Thermo Scientific SuperBlock Blocking Buffers for Blotting (Part No. 37517, 37537)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ReactivityHorseradish Peroxidase
Detection MethodColorimetric
FormLiquid
Product Line1-Step
Product TypeSubstrate
Quantity250 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°C. Product is shipped at ambient temperature.