GlutaMAX™ Supplement
Try new Pen-Strep antibiotic with GlutaMAX supplement, a stable alternative to L-glutamine that enhances cell health. Learn more >
GlutaMAX™ Supplement
Gibco™

GlutaMAX™ Supplement

Green features
GlutaMAX Supplement is an L-alanyl-L-glutamine dipeptide alternative to L-glutamine. It exhibits increased stability over L-glutamine leading to improved cellular health.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
35050061100 mL
3505007920 x 100 mL
Catalog number 35050061
Price (USD)
49.65
Online Exclusive
55.50
Save 5.85 (11%)
Each
Add to cart
Quantity:
100 mL
Price (USD)
49.65
Online Exclusive
55.50
Save 5.85 (11%)
Each
Add to cart

Gibco GlutaMAX Supplement is an L-alanyl-L-glutamine dipeptide alternative to L-glutamine that exhibits increased stability, leading to improved cell health. L-Glutamine is an amino acid required in cell culture for the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, and other amino acids, as well as in protein synthesis and glucose production. GlutaMAX Supplement is suitable for adherent and suspension culture of mammalian cells, with no adaptation required. It is formulated as a 200 mM L-alanyl-L-glutamine dipeptide in 0.85% NaCl.

Compared to L-glutamine, GlutaMAX Supplement:

  • Minimizes toxic ammonia build-up
  • Improves cell viability and growth
  • Remains stable across a wide range of temperatures

Minimizes toxic ammonia build-up

Unlike L-glutamine, GlutaMAX Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.

Improves cell viability and growth

Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX Supplement that is not possible with L-glutamine.

Remains stable across a wide range of temperatures

GlutaMAX Supplement is room-temperature stable and ready to use. It retains full functionality for 24 months at room temperature, 2 to 8°C, or -5 to -30°C for convenient storage and handling. GlutaMAX Supplement is even stable at 37°C for a week or more.

Specifications
Cell TypeMammalian Cells
Concentration100X
Culture TypeMammalian Cell Culture
For Use With (Application)Clinical Research, Translational Research
Green FeaturesSustainable packaging
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality430 to 490 mOsm/kg
Product LineGibco, GlutaMAX
Quantity100 mL
Shelf Life24 Months
Shipping ConditionRoom Temperature
SourceAnimal Origin-free
ClassificationAnimal Origin-free
FormLiquid
Product TypeGlutaMax
SterilitySterile-filtered
pH5 to 6
Unit SizeEach
Contents & Storage
Storage conditions: 15°C to 30°C
Alternate storage conditions: -5°C to -20°C and 2°C to 8°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture

Frequently asked questions (FAQs)

Why is GlutaMAX Supplement (Cat. Nos. 35050061, 35050079) recommended over L-glutamine when reconstituting Geltrex Reduced Growth Factor Basement Membrane Matrix?

GlutaMAX Supplement (Cat. No. 35050061, 35050079) is highly recommended to reconstitute Geltrex Reduced Growth Factor Basement Membrane Matrix because it offers higher stability than L-glutamine. The L-glutamine in Geltrex matrix comes in the form of the dipeptide L-alanyl-L-glutamine, which degrades slower than L-glutamine in cell culture. While GlutaMAX supplement is highly recommended, it is not required for Geltrex matrix–based culture systems.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Does Dynamis AGT Medium (Cat. Nos. A2617501, A2617502, A2617503, A2617504) require supplementation for use?

When using Dynamis AGT Medium (Cat. Nos. A2617501, A2617502, A2617503, A2617504) for culturing a CHO cell line other than the glutamine synthetase (GS) gene expression system (GS-CHO expression system), the addition of 2-8 mM L-glutamine or a similar amount of GlutaMAX Supplement (Cat. No. 35050079, 35050061) is recommended.

Note: We recommend that you supplement with glucose as needed so that glucose levels don’t fall below 2g/L in culture (see the product User Guide for more details).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do my cells need to adapt to GlutaMAX Supplement (Cat. No. 35050061, 35050079)?

Some cell lines may require an adaptation period, and the culture may lag in growth until it becomes accustomed to GlutaMAX Supplement (Cat. No. 3050061, 35050079). For serum-free media it is best to add L-glutamine along with GlutaMAX supplement. We don’t recommend that you use GlutaMAX supplement with insect cells such as Sf9.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between GlutaMAX Supplement (Cat. No. 35050061), CTS GlutaMAX-I Supplement (Cat. No. A1286001), and CTS GlutaMAX Supplement (Cat. No. A4737001)?

The formulation and manufacturing process are the same for all three products. The main difference is in the “intended use” statement. CTS GlutaMAX-I Supplement (Cat. No. A1286001) and CTS GlutaMAX Supplement (Cat. No. A4737001) have an intended use statement of “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals”. GlutaMAX Supplement (Cat. No. 35050061) has an intended use statement of “For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals”. The catalog number to use will depend on the specific application.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.