Nuclease-Free Water (not DEPC-Treated)
Invitrogen™

Nuclease-Free Water (not DEPC-Treated)

Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving.
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 x 50 mL
1 x 100 mL
1 x 500 mL
5 x 100 mL
1 x 1000 mL
4 x 1000 mL
Catalog number AM9932
Price (USD)
124.65
Online Exclusive
139.00
Save 14.35 (10%)
Each
Quantity:
1 x 1000 mL
Price (USD)
124.65
Online Exclusive
139.00
Save 14.35 (10%)
Each

Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving. This nuclease-free water is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Using Nuclease-free Water
Because this water is not DEPC-treated, it is recommended for applications for which residual DEPC might prove detrimental (e.g., oocyte microinjection).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingBottle
Product LineAmbion
Purification MethodAutoclaved, Membrane-Filtered
Recommended StorageStore at room temperature.
Shipping ConditionRoom Temperature
Treatment(s)Not DEPC-Treated
GradeMolecular Biology
Quantity1 x 1000 mL
Unit SizeEach

Frequently asked questions (FAQs)

What is the expiration date for Nuclease-Free Water (Cat. No. AM9937)?

This will be covered by our standard 12-month warranty as outlined in Section 8.1 of our Terms and Conditions of Sale. The 12-month warranty period begins from the date we ship the product to you. If your warranty has expired, the product may still perform as expected (when stored properly), we are just not able to guarantee or issue free-of-charge replacements in the case of poor performance.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.

I would like to order a TaqMan SNP Genotyping Assay. Does the kit include everything I need to perform my experiment or do I need to purchase reagents separately?

Each predesigned TaqMan SNP Genotyping Assay is delivered in a single tube consisting of two differentially labeled allele-specific TaqMan MGB (minor groove binding) probes and a pair of PCR primers. For more information, please see the following link: https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/real-time-pcr-applications/genetic-variation-analysis-using-real-time/snp-genotyping-with-real-time-pcr.html

Besides the TaqMan SNP Genotyping Assay, you would need to purchase a Genotyping Master Mix and nuclease-free water. A list of compatible master mixes can be found in the following link: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0009593_TaqManSNP_UG.pdf.

Find additional tips, troubleshooting help, and resources within our Gene Expression Analysis & Genotyping Support Center.