Nuclease-Free Water (not DEPC-Treated)
Actual product may vary
Nuclease-Free Water (not DEPC-Treated)
Nuclease-Free Water (not DEPC-Treated)
Invitrogen™

Nuclease-Free Water (not DEPC-Treated)

Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving.
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 x 50 mL
1 x 100 mL
1 x 500 mL
5 x 100 mL
1 x 1000 mL
4 x 1000 mL
Catalog number AM9938
Price (USD)
46.65
Online Exclusive
52.25
Save 5.60 (11%)
Each
Quantity:
1 x 100 mL
Price (USD)
46.65
Online Exclusive
52.25
Save 5.60 (11%)
Each

Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving. This nuclease-free water is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Using Nuclease-free Water
Because this water is not DEPC-treated, it is recommended for applications for which residual DEPC might prove detrimental (e.g., oocyte microinjection).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingBottle
Product LineAmbion
Purification MethodAutoclaved, Membrane-Filtered
Recommended StorageStore at room temperature.
Shipping ConditionRoom Temperature
Treatment(s)Not DEPC-Treated
GradeMolecular Biology
Quantity1 x 100 mL
pH6 to 8
Unit SizeEach

Frequently asked questions (FAQs)

I would like to order a TaqMan SNP Genotyping Assay. Does the kit include everything I need to perform my experiment or do I need to purchase reagents separately?

Each predesigned TaqMan SNP Genotyping Assay is delivered in a single tube consisting of two differentially labeled allele-specific TaqMan MGB (minor groove binding) probes and a pair of PCR primers. For more information, please see the following link: https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/real-time-pcr-applications/genetic-variation-analysis-using-real-time/snp-genotyping-with-real-time-pcr.html

Besides the TaqMan SNP Genotyping Assay, you would need to purchase a Genotyping Master Mix and nuclease-free water. A list of compatible master mixes can be found in the following link: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0009593_TaqManSNP_UG.pdf.

Find additional tips, troubleshooting help, and resources within our Gene Expression Analysis & Genotyping Support Center.