EASY-Spray™ HPLC Columns
Actual product may vary
EASY-Spray™ HPLC Columns
EASY-Spray™ HPLC Columns
EASY-Spray™ HPLC Columns
EASY-Spray™ HPLC Columns
Thermo Scientific™

EASY-Spray™ HPLC Columns

Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle SizePacking Material
ES907150 mm150 μm4 μmPSDVB (Polystyrene Divinylbenzene)
ES900150 mm75 μm3 μmSpherical, Fully Porous, Ultrapure Silica
ES901150 mm50 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES902250 mm75 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES903500 mm75 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES904150 mm75 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES905750 mm75 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES906150 mm150 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES911150 mm75 μm2.6 μmSpherical, Solid Core, Ultrapure Silica
Catalog number ES907
Price (USD)
1,725.46
Each
Length (Metric):
150 mm
Diameter (Metric):
150 μm
Particle Size:
4 μm
Packing Material:
PSDVB (Polystyrene Divinylbenzene)
Price (USD)
1,725.46
Each

Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.

EASY-Spray ̶ Fully integrated HPLC column/temperature/emitter design

  • Integrated fused-silica emitter delivers an exceptionally stable spray
  • Integrated column with temperature control provides maximum reliability and performance
  • A single nanoViper connection between the HPLC column and the MS ion source eliminates dead volumes
  • Thermo Scientific™ nanoViper™ fingertight fitting prevents overtightening and bad connections

Easy Spray Design Benefits

  • Uncompromised performance and ease-of-use in nano LC-MS
  • Ultra-sharp peaks
  • Outstanding peak capacity
  • Maximized peptide coverage
  • Outstanding column-to-column reproducibility and reliability
  • Outstanding peak capacity for comprehensive proteome characterization
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatEASY-Spray Capillary HPLC Column
Column TypeAnalytical HPLC Column
EndcappedN/A
For Use WithCapillary flow HPLC for sensitive protein, peptide and mAb separation
Max. Pressure8700 psi (600 bar)
Packing MaterialPSDVB (Polystyrene Divinylbenzene)
Particle ShapePolymeric
Stationary PhasePSDVB (Polystyrene Divinylbenzene)
Surface AreaN/A
Temperature60°C
USP TypeN/A
Diameter (Metric)150 μm
Length (Metric)150 mm
Particle Size4 μm
PhaseReversed Phase
Pore Size1500 Å
Product LineMAbPac
pH0 to 9
Unit SizeEach