Orion™ 8030cX Silica Analyzer
Thermo Scientific™
Orion™ 8030cX Silica Analyzer
Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeQuantity
8030CXSilica Analyzer1 Ea.
7070FLTSample Prefilter1 Ea.
803002Silica Standard Solution20 μg/L
803020Silica Standard Solution200 μg/L
803030Silica Standard Solution1000 μg/L
803040Silica Standard Solution4000 μg/L
8030CLCleaning Solution1 Ea.
8030MKMaintenance Kit1 Ea.
8010PCMProfibus Card1 Ea.
8030REXSilica Reagent Kit1 Ea.
Catalog number 8030CX
Price (USD)
-
Type:
Silica Analyzer
Quantity:
1 Ea.
Confidently perform silica analysis with the Thermo Scientific™ Orion™ 8030cX Silica Analyzer. Designed to provide accurate, effortless water analysis, the Orion 8030cX specializes in online measurement of reactive silica. Powered by advanced colorimetric technology, the Orion 8030cX delivers premium precision, greater productivity and enhanced durability. Supporting ultrapure and process water applications, the Orion 8030cX conveniently delivers precise, reliable measurement you can depend on. Unleash the possibilities and conquer the freedom of fast, confident decision making.
Confident Results
  • Low Detection Limit (0.5 ppb)
  • High Accuracy (≤1 ppb or 1% for range 0-500 ppb)
  • Patented Fluid Delivery & Quantification System
  • Reduced Phosphate Interferences
 Low Cost Ownership
  • 100 Days of Continuous Run Time (w/ 2L reagent and 15-minute measurement frequency)
  • Low Reagent Consumption (200 μL per measurement)
  • Separate Chemical Waste Collection (decreased disposal expenses)
 Easy to Use
  • Automated Calibration
  • Automated Validation & Verification
  • Color Touchscreen with Intuitive Interface
 Maximum Uptime
  • Automated Cleaning
  • Scheduled Measurements
  • Alert Notification System
  • Alarm System w/ Troubleshooting Guide
  • IP65 Certified (splash/ dust proof)
Markets
Silica monitoring for ultrapure water, demineralized water, deionized water, boiler water and steam condensate in:
  • Power Generation
  • Oil & Gas
  • Chemical Production
  • Semiconductor Manufacturing
  • Pulp & Paper
Applications
Monitoring and controlling silica levels in ultrapure and industrial process water to:
  • Prevent buildup on turbine blades, heat exchangers, boilers and more
  • Reduce costly and time-consuming cleaning procedures
  • Identify reverse osmosis (RO) breakdown and saturation of ion exchange resins
  • Maximize thermal efficiencies of steam passageways
  • Safeguard the health of critical plant assets by reducing potential for turbine vibration
Specifications
Accuracy≤ 1% or ± 1 μg/L (range 0-5000 ppb)≤ 5% (range 500-5000 ppb)
Analog OutputsTwo (2) isolated 4-20 mA, maximum load 900 Ω
Certifications/ComplianceSafety: cTUVus, CB, CE-LVD, UKCA, RCM EMC: FCC, CE-EMC, KC
Communication DetailsRS485, MODBUS RTU/TCP/IP, USB drive, PROFIBUS (optional)
Detection Limits0.5 μg/L (ppb)
Dimensions (L x W x H)406 mm x 678 mm x 320 mm (16.0 in x 26.7 in x 12.6 in)
Display Type7 in. color touchscreen
Electrical Requirements100 - 240 VAC, 100 W, 50/60 Hz
Enclosure RatingIP65 (splash and dust proof)
FrequencyMeasurement Frequency: Continuous or interval (10 - 720 minutes) Measurement Cycle: 10 minutes (fast mode) or 15 minutes (standard mode)
Max. Relative Humidity5% - 95% relative humidity, non-condensing
Mounting OptionsWall, panel or flat surface
Quantity1 Ea.
Range0-5000 μg/L (ppb)
Relay DetailsThree (3) relays, 8A @ 250 VAC
Repeatability≤ 1% or ± 0.6 μg/L (ppb)
Resolution0.01 μg/L for reading < 10 μg/L (ppb); 0.1 μg/L for reading ≥ 10 μg/L (ppb)
Sample Flow Rate50 - 300 mL/ minute
Sample InletConnector: Flow cell inlet OD (outer diameter); 8mm/ OD 6mm (1/4 inch)
Sample Pressure0.2 - 5.0 bar (2.9 - 72.5 psi)
Sample SupplyClean water with no particles over 0.1mm (prefilter 7070FLT can be used if large particles exist)
Sample Temperature0°C - 65°C (32°F - 149°F), no ice present
Temperature (English) Operating41°F - 113°F
Warranty1 year
Weight (English)Instrument Weight: 22.5 kg (49.5lbs) Shipping Weight: 36 kg (79.2 lbs)
Temperature (Metric) Operating5°C to 45°C
TypeSilica Analyzer
Unit SizeEach