Search
Built-in intelligence facilitates the full workflow from sample to results. Laboratories can be more reactive and profitable, with software that facilitates data driven decisions more efficiently.
Developed with scientific integrity, Qtegra ISDS Software delivers high quality, reliable data that labs can trust. Data transparency lets laboratory personnel focus on other tasks while ensuring the utmost confidence in scientific collaboration.
Data is the key to success for all laboratories. Qtegra ISDS Software can help labs excel at efficiently delivering consistent and relevant results with extensive tools to effectively acquire and manage data.
Laboratories can enjoy the familiarity of common software across multiple analytical systems while differentiating their offering through customizable and application specific workflows.
Qtegra ISDS Software is compatible with a variety of powerful analytical instruments used to analyze trace elements and elemental isotopes. Use the table below to see what features are available for your application.
| Supported | ICP-OES | Q-ICP-MS | HR-IRMS | MC-ICP-MS | Gas IRMS |
| Instruments | iCAP PRO ICP-OES | iCAP MX Series ICP-MS | Ultra HR-IRMS | Neoma MC-ICP-MS | DELTA Q IRMS and 253 Plus 10 kV IRMS with peripherals |
| Get Ready automated set-up | |||||
| Intuitive and guided workflow | |||||
| 21 CFR part 11 compliant environment | - | - | |||
| Autosampler | - | ||||
| Autodilution | - | - | |||
| Desolvation nebulizers | - | - | - | - | |
| Chromatography integration | - | Optional using ChromControl Plug-in | - | Optional using ChromControl Plug-in | |
| Single cell analysis | Optional using scQuant Plug-in | - | - | - | |
| Nanoparticle analysis | - | Optional using npQuant Plug-in | - | - | - |
| Laser plug-in | - | - | - | ||
| Semi-quant analysis | Axial only | - | - | - | - |
Download the Intelligent Scientific Data Solution Software product specifications for more details, including:
Trace elemental analysis and isotope analysis
Trace elemental analysis
Trace elemental analysis
Trace elemental analysis
Isotope analysis
* Required field