RadEye SPRD and SPRD-ER Personal Radiation Detector
Actual product may vary
RadEye SPRD and SPRD-ER Personal Radiation Detector
RadEye SPRD and SPRD-ER Personal Radiation Detector
RadEye SPRD and SPRD-ER Personal Radiation Detector
RadEye SPRD and SPRD-ER Personal Radiation Detector
Thermo Scientific™
RadEye SPRD and SPRD-ER Personal Radiation Detector
Experience extremely high radiation dose rate measurement and accuracy with the Thermo Scientific™ RadEye™ SPRD-ER Personal Radiation Detector.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
4250825RadEye SPRD-ER Personal Radiation Detector
425082541RadEye SPRD-ER Kit
425082542Radeye SPRD-ER Vehicle Kit
42508918RadEye SPRD-KIT
4250827RadEye SPRD Personal Radiation Detector
Catalog number 4250825
Price (USD)
-
Description:
RadEye SPRD-ER Personal Radiation Detector
Respond quickly and decisively to real radiation threats with the new standard in detection and interdiction of illicit radiation. The Thermo Scientific™ RadEye™ SPRD and SPRD-ER Personal Radiation Detectors are handheld radiation monitors that offer a unique combination of detector sensitivity and intelligent alarming to help ensure detection in the most challenging applications while eliminating nuisance alarms. Additionally, the RadEye SPRD and SPRD-ER radiation detectors identify and categorizes specific types of radiation or radioactive material, and can be configured to support your missions or user skills.
By providing significant detector sensitivity for search and find, combined with accurate high dose rate radiation measurement, the RadEye SPRD-ER is a suitable instrument for all users and may be the only radiation detector you need.
  • Faster detection of radiation at greater distances without increasing false alarms
  • Faster distinction between potential threats to public safety and non-threat sources (medical or natural)
  • Unprecedented count rate and dose rate accuracy across the full spectrum of keV – MeV
  • Simplified ID capability for novice to advanced users
  • Multiple modes of operation, easily configured to best align with existing radiation detection operating procedures
  • New Enhanced NBR identifies artificial radiation while minimizing false alarms
  • Extremely high dose rate measurement capabilities (RadEye SPRD-ER) of ≤1000 R/h (<10 Sv/h) supports users with one unit for search, find, radiation identification and response missions.
  • Sourceless detector gain stabilization improves radiation identification selectivity and accuracy across the spectrum in varying environmental conditions.
  • Stay on mission - detect radiation at greater distances
View our online training here.
Specifications
Battery Life> 170 hr. (alkaline)
Certifications/ComplianceANSI N42.32 PRD standard fully compliant; ANSI N42.48 SPRD standard fully compliant
CommunicationsWireless: Bluetooth 4.0 (Option)
Continuous Gain StabilizationSourceless spectral algorithm
DescriptionRadEye SPRD-ER Personal Radiation Detector
Detector Type1 Low Dose Rate Detector, 1 High Dose Rate Detector
Dimensions (L x W x H)4.1 x 2.6 x 1.6 in. (with rubber protective sleeve)
Dose Range10 nSv/hr to 10 Sv/hr (1 microR/hr to 1000 R/hr)
Drop Distance to Concrete1.5 m on concrete (with rubber sleeve)
Fast Gain Verification and AdjustmentLutetium Testadapter (< 10 nCi Lu-176)
MaterialCsI(Tl)
Natural Background RejectionAdvanced algorithms
Neutron Detection and VerificationPrompt gamma analysis
Operating Temperature (English)-4°F to +122°F
Operating Temperature (Metric)−20°C to +50°C
Sensitivity200 cps per μSv/h @ 662 keV
Typical ID Timeat 1μSv/h (100μR/h): < 3 min
Weight (English)0.429 lb. (including batteries and rubber sleeve)
Battery Type2 AAA alkaline or NiMH
Energy RangeSpectroscopic energy range: 20 keV - 3 MeV
Radiation TypeGamma, X-Ray plus neutrons via prompt gamma
Weight (Metric)0.195 kg (including batteries and rubber sleeve)
Unit SizeEach