SYBR Green dye-based master mixes for real-time PCR
When using dye-based chemistry for real-time PCR analysis, master mix choice matters. As the pioneers of SYBR Green dye quantification for qPCR, we have evolved our SYBR Green master mix formulations over many years into highly consistent reagents that deliver exceptional sensitivity and specificity on any qPCR instrument.
Compare SYBR Green vs TaqMan chemistries for qPCR analysis
SYBR Green master mix products
For optimal performance using SYBR green dye-based qPCR, we recommended using our latest formulation, PowerTrack SYBR Green Master Mix. For automated workflows that require 72-hour bench-top stability, we recommend PowerUP SYBR Green Master Mix.
PowerTrack SYBR Green Master Mix
PowerTrack SYBR Green Master Mix boosts reaction setup accuracy with an optional visual-tracking dye and simplifies primer optimization by accommodating a broad primer Tₘ range.
PowerUP SYBR Green Master Mix
PowerUp SYBR Green Master Mix features 72-hour benchtop stability with a dual hot-start mechanism for excellent specificity.
Compare PowerTrack and PowerUp SYBR Green master mixes
PowerTrack SYBR Green Master Mix
PowerUp SYBR Green Master Mix
Chemistry
2-step SYBR Green dye
2-step SYBR Green dye
Target input
cDNA
cDNA
Concentration
2X
2X
Polymerase Taq DNA Polymerase Dual-Lock Taq DNA Polymerase
Multiplexing and passive reference dye  With ROX: 1 target With ROX: 1 target
Cycling mode: run time
Fast: ~30 min
Standard: ~60 min
Fast: ~30 min
Standard: ~60 minutes
Sample tracking dye (optional) Yes No
Reaction benchtop stability 8 hours 72 hours
Carryover contamination control
With UNG
With UNG
Reverse transcriptase
(sold separately)
SuperScript IV Vilo Master Mix
High-Capacity RNA-to-cDNA™ Kit
Assay recommendations
Sequence detection primers
Sequence detection primers
Ordering information
Quick order below or visit product page
Visit product page
Other SYBR Green Master Mixes
Our legacy SYBR Green master mixes, including Power SYBR Green PCR Master MixSYBR Green PCR Master MixFast SYBR Green qPCR Master Mix, and SYBR Select Master Mix have long supported researchers with their qPCR analysis. However, for optimal results we recommend upgrading to our newest master mixes, which offer superior performance and compatibility with any qPCR instrument.
Master mix performance data
For reliably accurate qPCR data, our latest formulation, PowerTrack SYBR Green Master Mix, has been optimized to provide highly specific target amplficiation and excellent data reproducibility over a wide dynamic rage.
Figure 1. Highly specific amplification
Real-time PCR was performed using universal human reference (UHR) cDNA and primers targeting PGK1 (phosphoglycerate kinase 1), ARL1 (ADP-ribosylation factor-like protein 1), and SNF8 (vacuolar-sorting protein). Reactions were run in quadruplicate using master mixes from different suppliers. PowerTrack SYBR Green Master Mix generated a single melt curve in 100% of the reactions, indicating specific target amplification.
Figure 2. Excellent data reproducibility
PowerTrack SYBR Green Master Mix shows reproducibility over a wide dynamic range. Six assays (PGK1, ARL1,SNF8, DF, GAPDH, and Corf1 19) were run in quadruplicate with UHR cDNA generated from four different RT kits. Assays were run with a 6-fold dilution series and 400 nM primer concentration.
Instrument compatibility
Applied Biosystems SYBR Green master mixes can be used with any real-time PCR instrument, from any manufacturer. Refer to your master mix user guide for recommended experimental run conditions.
Ordering information
Quickly order PowerTrack SYBR Green Master Mix, PowerTrack SYBR Green Master Mix, reverse transcriptase reagents, and sequence detection primers in the tables below. To purchase other SYBR Green master mixes, visit the product page for ordering information.
PowerTrack SYBR Green Master Mix and reverse transcriptase
Catalog # Name Size Price Qty
A46012 Each
118.65
Online Exclusive
132.00 Save 13.35 (10%)
A46109 Each
511.65
Online Exclusive
568.00 Save 56.35 (10%)
A46110 Each
937.65
Online Exclusive
1,042.00 Save 104.35 (10%)
A46111 Each
2,245.65
Online Exclusive
2,495.00 Save 249.35 (10%)
A46112 Each
4,319.65
Online Exclusive
4,800.00 Save 480.35 (10%)
A46113 Each
4,306.65
Online Exclusive
4,785.00 Save 478.35 (10%)
11766050 Each
775.65
Online Exclusive
800.00 Save 24.35 (3%)
11756050 Each
737.65
Online Exclusive
760.00 Save 22.35 (3%)
11756500 Each
5,393.65
Online Exclusive
5,560.00 Save 166.35 (3%)
11766500 Each
5,810.65
Online Exclusive
5,990.00 Save 179.35 (3%)
PowerUp SYBR Green Master Mix and reverse transcriptase
Catalog # Name Size Price Qty
A25741 Each
116.65
Online Exclusive
130.00 Save 13.35 (10%)
A25742 Each
511.65
Online Exclusive
568.00 Save 56.35 (10%)
A25743 Each
4,274.65
Online Exclusive
4,750.00 Save 475.35 (10%)
A25778 Each
4,319.65
Online Exclusive
4,800.00 Save 480.35 (10%)
A25780 Each
541.65
Online Exclusive
602.00 Save 60.35 (10%)
A25776 Each
944.65
Online Exclusive
1,050.00 Save 105.35 (10%)
A25777 Each
2,245.65
Online Exclusive
2,495.00 Save 249.35 (10%)
4368814 Each
546.00
4374966 Each
888.00
4374967 Each
4,065.00
4368813 Each
2,080.00
Sequence detection primers
For Research Use Only. Not for use in diagnostic procedures.