Fast, accurate DNA, RNA, and protein quantification for a single sample
The Invitrogen Qubit 4 Fluorometer accurately and quickly measures the concentration of DNA, RNA, or protein in a single sample. It can also be used to assess RNA integrity and quality. The easy-to-use touchscreen menus make it easy to select and run the assays you need, with results displayed in just a few seconds.
Features
The Qubit 4 Fluorometer shares common features with the Qubit Flex model: It is accurate, specific, sensitive, simple, and fast; it requires as little as 1 µL of sample, even if highly diluted; it offers an onboard Reagent Calculator to help in preparing your working solution; it works with a broad range of Qubit Assays and applications; and it can measure raw fluorescence in Fluorometer mode.
In addition, data gathered in Fluorometer mode on the Qubit 4 Fluorometer can be used to design your own assay using the MyQubit assay design tool. This tool lets you create new assays for the Qubit Fluorometer in minutes. Simply enter your assay parameters into the online tool, and then save and upload the .qbt file to your Qubit Fluorometer using a USB drive.
Preconfigured MyQubit assays to quantify cholesterol, galactose, glucose, glutamic acid, peroxide, or sucrose are available. You can download these .qbt files to your computer and then upload them to your Qubit 4 Fluorometer using a USB drive. These are examples of the kinds of MyQubit assays you can create on your own.
Simple workflow
The user-friendly Qubit 4 Fluorometer delivers results using quick, simple assay procedures. The large, color touch screen allows you to easily navigate through the various options. Calculations and settings are automatically performed by the instrument.
Simple workflow
To quantify a sample with the Qubit 4 Fluorometer, select an assay, select the output, press Read tube, and view the results.
The Qubit 4 Fluorometer will save the data for up to 1,000 samples, and you can transfer data to a Thermo Fisher Connect Cloud account via WiFi, or to a computer via a USB drive or cable. In addition, the Qubit 4 integrated Reagent Calculator helps you determine the amount of dye and buffer required to prepare your samples and standards.
Reagent Calculator and results graph. Enter the number of samples and standards needed and the built-in Reagent Calculator will determine the amount of dye and buffer needed. The results can be viewed on a graph showing the standard curve.
Specifications
Attribute
Description
Physical characteristics
Instrument type
Benchtop fluorometer
Instrument dimensions
(W x L x H)
13.6 x 25 x 5.5 cm
5.4 x 10 x 2.2 in.
Weight
743 g; 2.1 lb
Operating power
100-240 VAC, 1.0 A/1.2 A
Frequency
50/60 Hz
Electrical input
12 VDC, 2.5 A
Installation site
Indoor use only
Operating temperature
10-40° C
Operating humidity
20-80% (non-condensing)
User interface
5.7-inch color touchscreen
Technical specifications
Dynamic range
5 orders of magnitude
Processing time
<5 seconds/sample
Light sources
Blue LED (peak ~470 nm)
Red LED (peak ~635 nm)
Excitation filters
Blue LED (430–495 nm)
Red LED (600–645 nm)
Emission filters
Green (510–580 nm)
Red (665–720 nm)
Detectors
Photodiodes: measurement capability from 300–1,000 nm
Calibration type
2- or 3-point standard
Sample chamber
Accommodates one Qubit Assay Tube
Tube type
Qubit Assay Tube
Warm-up time
<35 seconds
Sample data storage
1,000 samples
Hardware
Display
5.7 in. capacitive touchscreen with high-resolution color display
Data ports
USB cable port (type mini-B)
USB drive port (type A)
Networking capabilities
Transfer data directly to computer using a USB cable or wirelessly using the supplied Wi-Fi adaptor
Power supply
AC adapter with country-specific power cords
USB drive
4 GB
Ordering information
Starter kit comparison guide
Qubit 4 Fluorometer
Qubit 4 Quantitation Starter Kit
Qubit 4 NGS Starter Kit
Qubit 4 RNA IQ Starter Kit
Qubit 4 Protein Broad Range Starter Kit
Qubit 4 Fluorometer
Qubit 4 Fluorometer
Qubit 4 Fluorometer
Qubit 4 Fluorometer
Qubit 4 Fluorometer
1 international power adapter
1 international power adapter
1 international power adapter
1 international power adapter
1 international power adapter
Microfiber cloth
Microfiber cloth
Microfiber cloth
Microfiber cloth
Microfiber cloth
USB drive 
USB drive
USB drive
USB drive
USB drive 
WiFi dongle
WiFi dongle
WiFi dongle
WiFi dongle
WiFi dongle
Qubit Assay Tubes (500 tubes)
Qubit Assay Tubes (500 tubes)
Qubit Assay Tubes (500 tubes)
Qubit dsDNA HS (high sensitivity) Assay Kit (100 assays)
Qubit dsDNA HS (high sensitivity) Assay Kit (500 assays)
Qubit dsDNA BR (broad range) Assay Kit (100 assays)
Qubit RNA IQ (integrity and quality) Assay Kit (75 assays)
Qubit RNA IQ (integrity and quality) Assay Kit (275 assays)
Qubit RNA HS (high sensitivity) Assay Kit (100 assays)
Qubit RNA BR (broad range) Assay Kit (100 assays)
Qubit Protein BR Assay Kit (100 assays)
Qubit Protein BR Assay Kit (100 assays)
Qubit 4 System Verification Assay Kit (50 assays)
Q33238
Q33239
Q33240
Q33241
A51292
The Qubit 4 Fluorometer is designed to be used with Qubit Assay Tubes for best results.
Catalog # Name Size Price Qty
Q33238 Each
4,641.65
Online Exclusive
5,100.00 Save 458.35 (9%)
Q33239 Each
4,845.65
Online Exclusive
5,570.00 Save 724.35 (13%)
Q33240 Each
4,619.65
Online Exclusive
5,310.00 Save 690.35 (13%)
Q33241 Each
5,150.65
Online Exclusive
5,660.00 Save 509.35 (9%)
A51292 Each
5,010.65
Online Exclusive
5,230.00 Save 219.35 (4%)
Q32856 Each
128.00
Q33237 Each
100.00
For information about assays for the Qubit 4 Fluorometer, see the Assays tab.
For Research Use Only. Not for use in diagnostic procedures.