GeneStudioS5-chips-cartridge
Get a quote
Download specifications
Flexibility in throughput and workflow for a broad range of NGS applications
Chip type
Number of  reads
Read length (output) 
Ion GeneStudio S5 System
Ion GeneStudio S5 Plus System
Ion GeneStudio S5 Prime System
Turnaround time (sequencing run* and analysis time)
Ion 510 Chip
2–3M 200 bp (0.3–0.5 Gb)
4.5 hr
3 hr
3 hr
400 bp (0.6–1 Gb)
10.5 hr
5 hr
5 hr
Ion 520 Chip
4–6M 200 bp (0.6–1 Gb) 7.5 hr
3.5 hr
3 hr
400 bp (1.2–2 Gb)
12 hr
5.5 hr
5.5 hr
3–4M
600 bp (0.5–1.5 Gb)
12 hr
5.5 hr
5.5 hr
Ion 530 Chip
15–20M
200 bp (3–4 Gb)
10.5 hr
5 hr 4 hr
400 bp
(6-8 Gb)
21.5 hr 8 hr 6.5 hr
9–12M 600 bp (1.5–4.5 Gb)
21 hr
8 hr
7 hr
Ion 540 Chip
60–80M
200 bp (10–15 Gb)
19 hr
10 hr
6.5 hr
200 bp (20–30 Gb) 2 runs in 1 day
N/A
20 hr
10** hr
Ion 550 Chip
100–130M
200 bp (20–25 Gb)
N/A
11.5 hr
8.5 hr
200 bp (40–50 Gb) 2 runs in 1 day  N/A N/A
12** hr
† Expected output with >99% aligned/measured accuracy. Output dependent on read length and application.
* Sequencing run times are between 2.5 and 4 hrs.
** Analysis of first run occurs concurrently with the second sequencing run.