Varioskan LUX Multimode Microplate Reader
A delightfully easy to use multimode reader.
This multimode microplate reader combines simplicity with top-of-the-line performance. Designed for researchers with a wide variety of needs, the Thermo Scientific Varioskan LUX Multimode Microplate Reader offers features that help save time and reduce common errors in the lab—including automated dynamic range selection, which adjusts the optimal reading range based on signal intensities; and built-in safety controls, which alert you to potential errors before they happen.
Limited time offer
A 30-day money-back guarantee now through February 20th.
Use our configurator to build your Varioskan LUX system & get a quote
Features
Automatic dynamic range selection
Designed to automatically select the optimal reading range based on the signal intensity in each well. This feature saves time and reduces the risk of loss of sensitivity or over-saturation of signals.
Integrated gas control
The ability to control O2 and CO2 levels in your microplate reader replaces the need for constant handling of plates between the incubator and reader, and helps to ensure that cells remain alive throughout the experiment.
Smart safety controls
Automatic safety checks designed to alert you of potential problems before they happen. This decreases the potential of a range of experimental errors, from user mistakes to power outages.
Flexible wavelength selection
The instrument selects the measurement wavelength using filters or monochromators, depending on which is optimal for each measurement technology.
Available measurement technology configurations
All configurations available with top or top and bottom fluorescence intensity reading; none, one, or two dispensers; and optional integrated gas modules.
System configuration options
Absorbance and Fluorescence intensity
Absorbance, Fluorescence intensity, and Luminescence
Absorbance, Fluorescence intensity, Luminescence, and AlphaScreen
Absorbance, Fluorescence intensity, Luminescence, and Time-resolved fluorescence 
Absorbance, Fluorescence intensity, Luminescence, Time-resolved fluorescence, and AlphaScreen
Videos and demos
Specifications
Category
Attribute
Description
Absorbance
Plate types
6-384 well plates
Wavelength selection
Double monochromators
Wavelength range
200–1,000 nm
Light source
Xenon flash lamp
Read out range
0–6 Abs
Linear measurement range
0–4 Abs (96-well plate) at 450 nm, ± 2%
0–3 Abs (384-well plate) at 450 nm, ± 2%
Accuracy
0.003 Abs or ± 2%, at 200–399 nm (0–2 Abs)
0.003 Abs or ± 1%, at 400–1,000 nm (0–3 Abs)
Precision
SD <0.001 Abs or CV <0.5%, at 450 nm (0–3 Abs)
Fluorescence Intensity
Plate types
6 – 1536-well plates (top reading)
6 – 384-well plates (bottom reading)
Wavelength selection
Double excitation and emission monochromators
Excitation wavelength range
200–822 nm
Emission wavelength range
270–840 nm
Light source
Xenon flash lamp
Sensitivity
Top reading: <0.4 fmol fluorescein/well (black 384-well plate)
Bottom reading: <4 fmol fluorescein/well, (clear bottom black 384 square well plate)
Dynamic range
Top reading >6 decades
Bottom reading >5.5 decades
Time-resolved fluorescence
Plate types
6–1,536 well plates
Wavelength selection
Filters (spectral scanning with double excitation and emission monochromators)
Excitation wavelength range
Fixed to 334 nm (spectral scanning 200–840 nm)
Emission wavelength range
400–670 nm (spectral scanning 270–840 nm)
Light source
Xenon flash lamp
Sensitivity
<1 amol Eu/well (white low volume 384 well plate)
Dynamic range
>6 decades
Luminescence
Plate types
6–1536 well plates (spectral scanning 6-384 well plates)
Wavelength selection
Direct or filters (spectral scanning with double monochromators)
Wavelength range
360–670 nm
Sensitivity
<7 amol ATP/well (white 384-well plate)
Dynamic range
>7 decades
Alpha
Plate types
6-1536 well plates
Wavelength selection
Filters
Excitation wavelength range
Fixed to 680 nm
Emission wavelength range
400–660 nm
Light source
LED
Sensitivity
<100 amol phosphotyrosine/well (white 384-well plate)
Dispensing
Plate types
6-384 well plates
No. of dispensers
None, one or two
Syringe size
1 mL 
Dispensing volume
2–5,000 μL, in 1 µL increments
Accuracy
<1 μL with 50 μL
Precision
<1 μL with 50 μL
Dead volume
Reagent loss <100 μL, Total tubing volume <800 μL
Incubator and Shaker
Temperature range
From ambient + 4 °C to 45 °C
Shaking type
Orbital
Integrated Gas Module
Measurement chamber O2 concentration accuracy
±1.0% (37 °C, 1% O2)
Measurement chamber CO2 concentration accuracy
±1.0%. (37 °C, 5% CO2)
Measurement chamber O2 concentration
1-21%
Measurement chamber CO2 concentration
0.1-15%
Gas concentration recovery time
10 minutes
General Features
Measurement modes
Endpoint, kinetic, spectra, multipoint and kinetic spectra
Measurement speed
Reads a 96-well plate in 15 s, a 384-well plate in
45 s, and a 1,536-well plate in 135 s (minimum times)
Interface
PC software (SkanIt software)
Dimensions (D x W x H)
58 x 53 x 51 cm (23 x 21 x 20 in)
Weight
53 - 60 kg, (117-132 lb), depending on configuration