For Clinicians
Resources for Medical Professionals  
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
  Food  
  Seasonal  
  Indoor  
  Allergic Asthma  
Is your patient a candidate for specific IgE testing?
Find out 
Get detailed information on whole allergens and allergen components. 
Allergen Encyclopedia 
Ready to test a patient?
On-Demand Education 
Cat Dander
Allergy Facts, Symptoms, and Treatment
Up to 60 percent of European and U.S. households contain pets, and cats and dogs are the two most popular varieties.1 Unfortunately, allergies to cats affect 7 to 25 percent of the population.2 These allergic individuals, then, are sensitized to one or more allergens that are present in cat saliva, urine, and dander. However, these allergens often stick to animal hair and dander and are easily dispersed; plus, they can become airborne and linger suspended in air.1,3 Those allergic to cat allergens may experience symptoms when allergens are inhaled (either via direct or indirect contact with a cat) and when allergenic proteins are delivered via cat bite.1 Regardless of hypoallergenic claims, all cats produce a key allergic protein, with males having a higher concentration.4
Where is cat dander found?
While cat allergens are present in the animal's saliva, urine, and dander, they can become airborne and linger suspended in air for long periods.1,3 Plus, they can end up in myriad environments including those with or without cats, such as schools, day-care centers, households, and transportation facilities.1 These allergens also can cling to many surfaces, such as bedding, clothing, and upholstered furniture, making them ubiquitous.1,7 And unfortunately, cat allergens can cause year-round symptoms and may remain in an environment for months without degrading.4
Prepare for your next visit with your healthcare provider. 
Fill out our symptom tracker
Learn about specific IgE allergy testing options.
Read more
Are there other allergens I could be sensitized to?*
Some people with a cat allergy may also experience symptoms when exposed to dander, saliva, and urine from other mammalian animals, such as dogs, guinea pigs, horses, pigs, chickens, sheep, goats, rabbits, and hamsters. In addition, ingestion of foods such as milk, eggs, beef, and pork (as seen in pork-cat syndrome) may also elicit symptoms. This is called cross-reactivity and occurs when your body's immune system identifies the proteins, or components, in different substances as being structurally similar or biologically related, thus triggering a response.1
Knowing the proteins, or components, within each allergen that are triggering your symptoms can help guide your management plan. With that in mind, and based on your symptom history, your healthcare provider may suggest something called a specific IgE component test, which can help reveal the risk profile you belong to. Results from this test can also help your healthcare provider decide if immunotherapy may reduce your symptoms.1
Already have your specific IgE component test results?
Your component test results will include the name of the components (a series of letters and numbers). Your healthcare provider will likely review the results with you, but here you'll find an at-a-glance breakdown you can use as a reference. Simply match the component names to the list below to see what they mean in terms of symptom management.1,8
rFel d 1     
  • Marker for risk of severe asthma.
  • Cat allergen immunotherapy may be an option.
  • Indicates that symptoms may be caused specifically by cat.
rFel d 2, rFel d 4         
  • Cats or other mammalian pets, e.g., dogs, horses, and mice, may cause symptoms due to cross-reactivity.
  • Ingestion of milk, beef, and pork (pork-cat syndrome) may elicit symptoms due to cross-reactivity. Cooked milk and beef may be tolerated.
  • High levels of IgE to these proteins are acociated with atopic dermatitis in cat-allergic children.
rFel d 7     
  • Dogs may cause symptoms due to cross-reactivity.
Test results should be interpreted by your healthcare provider in the context of your clinical history. Final diagnosis and decision on further management is made by your healthcare provider.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
How do I manage my allergy?
If you are allergic to cats, your healthcare provider may recommend a plan that includes the following.1,3,5,7-9,10
The best way to reduce exposure is to keep cats out of your home and to avoid visiting environments with cats. However, if you can't bear to give up an existing pet, the following strategies may help reduce allergen exposure.
  • Prevent the cat from entering the bedroom, keep the door closed, and clean the bedroom aggressively. Also consider using a HEPA air cleaner in the bedroom.
  • Remove wall-to-wall carpet and scrub the walls and woodwork. Keep surfaces throughout the home clean and uncluttered. Bare floors and walls are best.
  • If you must have carpet, select one with a low pile and steam clean it frequently. Better yet, use throw rugs and wash them in hot water.
  • Use a vacuum with a certified asthma and allergy friendly filter, and wear a dust mask when vacuuming.
  • Refrain from touching your eyes after handling a cat, and wash your hands immediately. Also change your clothes if possible.
  • Add an air cleaner combined with a certified asthma and allergy friendly filter to central heating and air conditioning systems to help remove allergens from the air.
  • Remove pillows and other items that may act as reservoirs.
  • Wash the pet and pet bedding frequently.
Your healthcare provider may direct you to employ one of the following therapies to improve your allergy symptoms:
  • Antihistamines are commonly used to reduce symptoms such as sneezing, itching, and runny nose.
  • Nasal corticosteroids are used to reduce swelling in the nose and block allergic reactions.
  • Decongestants can be employed to relieve stuffiness.
  • Allergen immunotherapy, as directed by your healthcare provider, may help develop tolerance to cat dander exposure.
If you're with someone who's having an allergic reaction and shows signs of shock, act fast. Look for pale, cool, and clammy skin; a weak, rapid pulse; trouble breathing; confusion; and loss of consciousness. Do the following immediately:
  • Call local emergency services.
  • Ensure the person is lying down and elevate his or her legs.
  • Administer epinephrine immediately for any obvious signs of a potentially severe systemic reaction.
  • Check the person’s pulse and breathing and administer CPR or other first-aid measures if necessary.
Looking for more allergy info and management tips?
Visit the Living with Allergies section
Common Symptoms
Symptoms of cat allergy can include:5
  • Sneezing
  • Runny nose
  • Itchy, red, or water eyes
  • Nasal congestion
  • Itchy nose, mouth, or throat
  • Postnasal drip
  • Cough
  • Facial pressure and pain
  • Frequent awakening form sleep
  • Swollen, blue-colored skin under eyes
If you have asthma, you may also experience symptoms such as:5
  • Difficulty breathing
  • Chest tightness or pain
  • Audible wheezing or whistling when exhaling
  • Trouble sleeping caused by shortness of breath, coughing, or wheezing
In addition, some people may also experience symptoms of allergic dermatitis after direct contact with a cat. These symptoms may include:5
  • Raised red patches of skin (hives)
  • Eczema
  • Itchy skin
Although most pet-allergy symptoms occur within minutes of exposure, symptoms in some allergic individuals build over time and become most severe eight to 12 hours later.6
How do I know if I'm allergic?*
Together with your symptom history, skin-prick testing or specific IgE blood testing can help determine if you are allergic to a particular allergen. If you are diagnosed with an allergy, your healthcare provider will work with you to create a management plan.
*These products may not be approved for clinical use in your country. Please work with your healthcare provider to understand availability.
Is there a risk for a severe event?
Sensitization to furry animals is common and a risk factor for the development of allergic rhinitis and asthma.1 In fact, high levels of IgE for cat, dog, and horse allergen components are markers for severe asthma. Sensitization to all three animals at the same time is also a risk marker of severe asthma.1,8 Also note that some animal bites are capable of producing anaphylaxis, but it's unusual for a cat bite to evoke this reaction.1
  1. EAACI, et al. Molecular allergology user's guide. Pediatric Allergy Immunol. 2016 May;27 Suppl 23:1-250. do: 10.1111/pai.12563. PMID: 27288833. (123-130 p.) Available from: http://www.eaaci.org/documents/Molecular_Allergology-web.pdf.
  2. Satyaraj E, Wedner HJ, Bousquet J. Keep the cat, change the care pathway: A transformational approach to managing Fel d 1, the major cat allergen. Allergy. 2019 Oct;74 Suppl 107(Suppl 107):5-17. doi: 10.1111/all.14013. PMID: 31498459; PMCID: PMC7156987. Available from: https://onlinelibrary.wiley.com/doi/full/10.1111/all.14013.
  3. Asthma and Allergy Foundation of America [Internet]. Arlington, VA: Asthma and Allergy Foundation of America; 2015 Oct. Available from: https://www.aafa.org/pet-dog-cat-allergies.
  4. Baxi SN, Phipatanakul W. The role of allergen exposure and avoidance in asthma. Adolesc Med State Art Rev. 2010 Apr;21(1):57-71, viii-ix. PMID: 20568555; PMCID: PMC2975603. Available from: https://www.ncbi.nlm.nih.gov/pmc/articles/PMC2975603.
  5. Mayo Clinic [Internet]. Rochester, MN: Mayo Foundation for Medical Education and Research; 2019 May 14. Available from: https://www.mayoclinic.org/diseases-conditions/pet-allergy/symptoms-causes/syc-20352192.
  6. American Academy of Allergy, Asthma & Immunology [Internet]. Milwaukee WI, American Academy of Allergy, Asthma & Immunology. 2020 Sep 28. Available from: https://www.aaaai.org/conditions-and-treatments/library/allergy-library/indoor-allergens.
  7. Harvard Health Publishing [Internet]. Boston, MA: Harvard Medical School; 2009 Oct. Available from: https://www.health.harvard.edu/press_releases/tips-to-control-pet-allergies.
  8. Dávila I, Domínguez-Ortega J, Navarro-Pulido A, Alonso A, Antolín-Amerigo D, González-Mancebo E, Martín-García C, Núñez-Acevedo B, Prior N, Reche M, Rosado A, Ruiz-Hornillos J, Sánchez MC, Torrecillas M. Consensus document on dog and cat allergy. Allergy. 2018 Jun;73(6):1206-1222. doi: 10.1111/all.13391. Epub 2018 Feb 13. PMID: 29318625.
  9. Mayo Clinic [Internet]. Rochester, MN: Mayo Foundation for Medical Education and Research; 2019 May 14. Available from: https://www.mayoclinic.org/diseases-conditions/pet-allergy/diagnosis-treatment/drc-20352198.
  10. Mayo Clinic [Internet]. Rochester, MN: Mayo Foundation for Medical Education and Research; 2019 Sep 14. Available from: https://www.mayoclinic.org/diseases-conditions/anaphylaxis/diagnosis-treatment/drc-20351474#:~:text=During%20an%20anaphylactic%20attack%2C%20you,Oxygen%2C%20to%20help%20you%20breathe.