桑樹 過敏原基本資料、症狀和治療
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
Learn more about common allergic diseases, symptoms, management paradigms, and testing considerations.
Is your patient a candidate for specific IgE testing?
Get detailed information on whole allergens and allergen components.
Ready to test a patient?
桑樹因其果實而被種植,也是蠶的重要食物來源,有三個主要品種:紅桑樹、白桑樹和黑桑樹。1 桑樹花粉透過風吹傳播,在許多地方是重要的呼吸道過敏原,桑樹是可能具有高度致敏性的大量授粉植物。2,3,4
紅桑樹生長在北美,白桑樹則原生於亞洲並在歐洲南部也有種植。在這三種常見品種中,黑桑樹最開始生長在西亞,然後傳入歐洲和北美。1
許多桑樹過敏患者在接觸其他過敏原 (如木本、雜草或草本花粉) 時也會出現症狀,因此很難確定是哪種花粉引起症狀,尤其是花粉季節有重疊時。這稱為交叉反應,當人體的免疫系統認定不同物質中的蛋白質或成分在結構上相似或生物學上相關,就會發生交叉反應,從而引發免疫反應。9 其他可能引起與桑樹花粉過敏相關的呼吸道過敏原包含樺樹、牆草屬植物和橄欖樹花粉。10
*在您的國家/地區,這些產品可能並未獲准用於臨床用途。請與醫護人員溝通,以了解其可用性。
過敏性鼻炎的控制方法包括避開相關過敏原、對症治療和過敏原免疫療法。6-8
桑樹過敏症狀可能與許多其他花粉過敏相似,可能包括:5.6
如果您對桑樹過敏並患有氣喘,則木本花粉可能會觸發或加劇氣喘症狀,例如咳嗽和喘鳴。5.6
結合您的症狀史,皮膚點刺檢測或血清特異性 IgE 檢測有助於確定您是否對特定過敏原敏感。如果您確診患有過敏症,醫護人員將與您一起制定控制計畫。
*在您的國家/地區,這些產品可能並未獲准用於臨床用途。請與醫護人員溝通,以了解其可用性。
雖然樹木花粉常見於春季,但桑樹在春冬兩季都會授粉。2,5