  • Applications
    • Bioprocédés
    • Culture cellulaire et transfection
    • Thérapie cellulaire et génétique
    • Chromatographie
    • Tests moléculaires
    • Solutions numériques
    • Extraction et analyse d’ADN et d’ARN
    • Spectroscopie, analyse élémentaire et isotopique
    • Voir toutes les applications et techniques
  • Services
    • Services CDMO et CRO à 360°
    • Services CDMO
    • Services CRO
    • Services personnalisés
    • Services financiers et leasing
    • Services pour les instruments
    • Informatique de laboratoire
    • Offres commerciales et OEM
    • Services de formation
    • Unity Lab Services
    • Voir tous les services
  • Aide et assistance
    • S’inscrire et créer un compte
    • Comment commander
    • Assistance instruments
    • Centres d’assistance technique
    • Centres d’apprentissage
    • Consultez toutes les rubriques d'aide et d'assistance
  • Populaire
    • TaqMan Real-Time PCR Assays
      Reaction PCR en temps réel TaqMan
    • Antibodies
      Anticorps
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt - Synthèse Gène
    • Cell Culture Plastics
      Plastiques culture cellulaire
  • Nos clients
    • Biotechnologie
    • Industrie biopharmaceutique
    • CDMO
    • Diagnostics de laboratoire
    • Sciences industrielles et appliquées associées
  • Promotions
  • Contactez-nous
  • Commande rapide
  • Suivi et statut de la commande
  • Documents et certificats
Thermo Fisher Scientific Logo
Search
Rechercher
Search button
          • Statut des commandes
          • Commande rapide
          • Promos
          • Se connecter
            Se connecter
            Vous n’avez pas de compte? Créer votre compte
            • Mon compte
            • Commandes
            • Connect: lab, données, apps
            • Produits personnalisés et projets
            • Services Central
          • Primary Antibodies ›
          • CD4 Antibodies
          Invitrogen
          CD4 Monoclonal Antibody (RM4-4), FITC, eBioscience™
          Advanced Verification
          7 Published Figures
          30 References
          View all (691) CD4 antibodies
          Tech Support
          Cite CD4 Monoclonal Antibody (RM4-4), FITC, eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (7)
          • Advanced Verification (1)
          CD4 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD4 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 9
          CD4 Antibody (11-0043-85) in Flow
          C57BL/6 mouse splenocytes were stained with Rat IgG2b kappa Isotype Control, FITC (Product # 11-4031-82) (blue histogram) or CD4 Monoclonal Antibody, FITC (purple histogram). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD4 Antibody in Flow Cytometry (Flow)
          CD4 Antibody in Flow Cytometry (Flow)
          CD4 Antibody in Flow Cytometry (Flow)
          CD4 Antibody in Flow Cytometry (Flow)
          CD4 Antibody in Flow Cytometry (Flow)
          CD4 Antibody in Flow Cytometry (Flow)
          CD4 Antibody in Flow Cytometry (Flow)
          CD4 Antibody in Immunohistochemistry (IHC)
          CD4 Antibody
          Product Details
          11-0043-85
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          -
          View 1 publication 1 publication
          Immunocytochemistry (ICC/IF)
          -
          View 2 publications 2 publications
          Flow Cytometry (Flow)
          0.125 µg/test
          View 25 publications 25 publications
          Miscellaneous PubMed (Misc)
          -
          View 2 publications 2 publications
          Product Specifications
          Species Reactivity
          Mouse
          Published species
          Human, Mouse
          Host/Isotype
          Rat / IgG2b, kappa
          Recommended Isotype Control
          Rat IgG2b kappa Isotype Control (eB149/10H5), FITC, eBioscience™
          Class
          Monoclonal
          Type
          Antibody
          Clone
          RM4-4
          Conjugate
          FITC FITC FITC
          • APC 
          • Biotin 
          • PE 
          • PE-eFluor 610
          • Request custom conjugation
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          500 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2
          Contains
          0.09% sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_464901
          Product Specific Information
          Description: The RM4-4 monoclonal antibody reacts with the mouse CD4 molecule, a 55 kDa cell surface receptor expressed by the majority of thymocytes, a subpopulation of mature T cells and dendritic cells. CD4 binds to MHC class II on the surface of antigen presenting cells and plays an important role both in T cell development and in optimal functioning of mature T cells. In T cells, CD4 associates with the protein tyrosine kinase lck through its cytoplasmic tail. Binding of RM4-4 does not block binding of the CD4 monoclonal antibodies RM4-5 or GK1.5.
          Applications Reported: This RM4-4 antibody has been reported for use in flow cytometric analysis.
          Applications Tested: This RM4-4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: CD4; CD4 antigen p55; cd4a; fCD4; Leu-3; T-cell differentiation antigen L3T4; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4; T-cell surface glycoprotein CD4 precursor (T-cell surface antigen T4/Leu-3) (T-cell differentiation antigen L3T4)
          View more View less
          Gene Aliases: Cd4; L3T4; Ly-4
          View more View less
          UniProt ID: (Mouse) P06332
          View more View less
          Entrez Gene ID: (Mouse) 12504
          View more View less
          Function(s)
          glycoprotein binding protein binding zinc ion binding coreceptor activity enzyme binding protein kinase binding protein homodimerization activity
          Process(es)
          cytokine production immune system process induction by virus of host cell-cell fusion immune response cell adhesion cell surface receptor signaling pathway T cell differentiation maintenance of protein location in cell T cell activation T cell selection protein palmitoleylation positive regulation of protein kinase activity positive regulation of peptidyl-tyrosine phosphorylation defense response to Gram-negative bacterium positive regulation of calcium-mediated signaling regulation of T cell activation positive regulation of T cell activation
          Contact tech support
          Commander Plus Icon Minus Icon
          • Statut des commandes
          • Assistance commandes
          • Commande rapide
          • Centre d’approvisionnement
          • Approvisionnement en ligne
          Assistance Plus Icon Minus Icon
          • Aide et assistance
          • Contactez-nous
          • Centres d’assistance technique
          • Documents et certificats
          • Signaler un problème sur le site
          Ressources Plus Icon Minus Icon
          • Centres d’apprentissage
          • Promotions
          • Événements et séminaires en ligne
          • Réseaux sociaux
          À propos de Thermo Fisher Plus Icon Minus Icon
          • À propos de nous À propos de nous
          • Emplois Emplois
          • Investisseurs Investisseurs
          • Actualités Actualités
          • Responsabilité sociale Responsabilité sociale
          • Marques commerciales
          • Politiques et notices d’information
          Notre portfolio Plus Icon Minus Icon
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline