    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PU.1 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          PU.1 Monoclonal Antibody (phpu13), eBioscience™
          Advanced Verification
          1 Reference
          View all (32) PU.1 antibodies
          Tech Support
          Cite PU.1 Monoclonal Antibody (phpu13), eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          PU.1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          PU.1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          PU.1 Antibody (14-9819-80) in IHC (P)
          Immunohistochemistry on formalin-fixed paraffin embedded human infiltrating ductal carcinoma, using 10 µg/mL of mouse IgG1 isotype control (left) or 10 µg/mL Anti-Human/Mouse PU-1 Purified (right) followed by biotinylated anti-mouse IgG, and DAB visualization.Nuclei are counterstained with hematoxylin. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PU.1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PU.1 Antibody in Western Blot (WB)
          PU.1 Antibody
          Product Details
          14-9819-80
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (IHC)
          -
          View 1 publication 1 publication
          Immunohistochemistry (Paraffin) (IHC (P))
          10 µg/mL
          -
          Immunocytochemistry (ICC/IF)
          Assay-Dependent
          -
          Product Specifications
          Published species
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          phpu13
          Immunogen
          E. coli-derived protein, without DNA domain
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          25 µg
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          RRID
          AB_11150963
          Product Specific Information
          Description: The monoclonal phpu13 recognizes human and mouse PU.1, a member of the ets family of transcription factors. This 42 kDa protein was originally identified as Spi-1, a proto-oncogene, for its role in Friend virus-induced murine erythroleukemia. Expression of PU.1 plays a critical role in lymphocyte and myeloid development. Knock-out mice are neonatal lethal and lack myeloid, T and B cells. In addition erythroid development is also altered. Protein expression levels help guarantee proper development; higher levels in myeloid cells compared to lymphocytes. PU.1 controls transcription of several proteins including CD11b, CD18, Gata2, IRF4, CD20, CD10, and CD79.
          Disruption of PU.1 expression may play a role in several specific leukemias; such as B-chronic lymphocytic leukemia, mantle cell lymphoma, follicular lymphoma, marginal zone lymphoma, Burkitt lymphoma, diffuse large cell lymphoma, diffuse large B-cell lymphoma, T-cell rich B-cell lymphoma, and nodular lymphocyte predominant Hodgkin.
          Applications Reported: This phpu13 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections and immunocytochemistry.
          Applications Tested: This phpu13 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded tissue using IHC Antigen Retrieval Solution - Low pH (10X) (Product # 00-4955-58). This can be used at less than or equal to 10 µg/mL. It is recommeded that the antibody be carefully titrated for optimal performance in the assay of interest.
          Purity: Greater than 90%, as determined by SDS-PAGE.
          Aggregation: Less than 10%, as determined by HPLC.
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          This gene encodes an ETS-domain transcription factor that activates gene expression during myeloid and B-lymphoid cell development. The nuclear protein binds to a purine-rich sequence known as the PU-box found near the promoters of target genes, and regulates their expression in coordination with other transcription factors and cofactors. The protein can also regulate alternative splicing of target genes. Multiple transcript variants encoding different isoforms have been found for this gene.
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline