eBioscience™ IHC Antigen Retrieval Solution - Low pH (10X)
Invitrogen™

eBioscience™ IHC Antigen Retrieval Solution - Low pH (10X)

The IHC Antigen Retrieval Solution- Low pH (10X) is designed for use during the heat induced epitope retrieval (HIER) stepRead more
Have Questions?
Catalog NumberQuantity
00-4955-58500 mL
Catalog number 00-4955-58
Price (TWD)
7,350.00
Online offer
Ends: 31-Dec-2026
10,500.00
Save 3,150.00 (30%)
Each
Quantity:
500 mL
Price (TWD)
7,350.00
Online offer
Ends: 31-Dec-2026
10,500.00
Save 3,150.00 (30%)
Each
The IHC Antigen Retrieval Solution- Low pH (10X) is designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. The use of this buffer in combination with heat (often by microwave, water bath, or pressure cooker) has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue. This buffer is supplied as a 10X stock solution. Dilution of this buffer to 1X using reagent grade water results in a 10 mM Sodium Citrate solution, pH 6.0.

Reported Application
Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product TypeIHC Antigen Retrieval Solution
For Use With (Application)Immunohistochemistry (IHC)
Product LineeBioscience
Quantity500 mL
Unit SizeEach

Citations & References (4)

Citations & References
Abstract
Pterostilbene reduces endothelial cell injury in vascular arterial walls by regulating the Nrf2-mediated AMPK/STAT3 pathway in an atherosclerosis rat model.
Authors:Tang T, Duan Z, Xu J, Liang J, Zhang S, Zhang H, Zhang X, Wang Y
Journal:Exp Ther Med
PubMed ID:31853271
'Endothelial cell injury in vascular arterial walls is a hallmark of atherosclerosis. Pterostilbene (Pts) has been shown to have an anti-oxidative and anti-apoptotic effect in numerous diseases via regulation of intracellular metabolism. The purpose of this study was to investigate the protective effect and possible mechanism of Pts against endothelial ... More
Puerarin inhibits apoptosis and inflammation in myocardial cells via PPARa expression in rats with chronic heart failure.
Authors:He L, Wang T, Chen BW, Lu FM, Xu J
Journal:Exp Ther Med
PubMed ID:31602208
'Chronic heart failure affects myocardial energy metabolism and cardiac function. Puerarin has been reported to improve cardiac function through regulation of energy metabolism in mice with myocardial infarction. The aim of the current study was to determine whether puerarin can improve body weight and reduce inflammation and apoptosis in rats ... More
Contrast-enhanced computed tomography combined with Chitosan-Fe
Authors:Gai J, Gao Z, Song L, Xu Y, Liu W, Zhao C
Journal:Exp Ther Med
PubMed ID:29805549
Esophageal cancer is a malignant tumor with a relatively high invasiveness, metastatic potential and worldwide incidence among human cancers. The majority of patients with esophageal cancer are diagnosed in a late tumor stage due to a lack of advanced and sensitive protocols for the diagnosis of patients with early-stage esophageal ... More
Management of primary pterygium with intra-lesional injection of 5 flurouracil and bevacizumab (Avastin).
Authors:Ghoz N, Britton J, Ross AR, Mohammed I, Hogan E, Said DG, Dua HS
Journal:Eye (Lond)
PubMed ID:31217579
To assess the efficacy of combined 5FU and Avastin injections in the treatment of primary pterygium METHODS: Sixteen eyes with primary pterygium received intralesional 5 fluorouracil and Avastin (2.5-5?mg) injections every 2 weeks for a maximum of five injections. Fourteen eyes of 14 patients received five injections, one eye received ... More