    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • IL-5 Antibodies
          Invitrogen
          IL-5 Monoclonal Antibody (TRFK5), Functional Grade, eBioscience™
          8 Published Figures
          6 References
          View all (53) IL-5 antibodies
          Tech Support
          Cite IL-5 Monoclonal Antibody (TRFK5), Functional Grade, eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (8)
          IL-5 Antibody in Immunostaining (IS)
          Group 53 Created with Sketch.
          IL-5 Antibody in Immunostaining (IS)
          Group 53 Created with Sketch.
          FIGURE: 1 / 9
          IL-5 Antibody (16-7052-85) in IS
          Naive mouse splenocytes (SJL/J strain) were stimulated with 2 µg/mL ConA in HL-1 media for 48 hours (right well) in a Mouse IL-5 ELISPOT assay plate. Left well is medium alone control. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          IL-5 Antibody in Immunostaining (IS)
          IL-5 Antibody in Flow Cytometry (Flow)
          IL-5 Antibody in Flow Cytometry (Flow)
          IL-5 Antibody in Flow Cytometry (Flow)
          IL-5 Antibody in Flow Cytometry (Flow)
          IL-5 Antibody in ELISA (ELISA)
          IL-5 Antibody in ELISA (ELISA)
          IL-5 Antibody in Radioimmune assays (RIA)
          IL-5 Antibody in Neutralization (Neu)
          Product Details
          16-7052-85
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          Assay-Dependent
          -
          Flow Cytometry (Flow)
          -
          View 2 publications 2 publications
          ELISA (ELISA)
          -
          View 1 publication 1 publication
          Neutralization (Neu)
          Assay-Dependent
          View 1 publication 1 publication
          Functional Assay (Functional)
          Assay-Dependent
          View 1 publication 1 publication
          Radioimmune Assays (RIA)
          -
          View 1 publication 1 publication
          Immunostaining (IS)
          0.06 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Published species
          Mouse
          Host/Isotype
          Rat / IgG1
          Recommended Isotype Control
          Rat IgG1 kappa Isotype Control (eBRG1), Functional Grade, eBioscience™
          Class
          Monoclonal
          Type
          Antibody
          Clone
          TRFK5
          Conjugate
          Functional Grade Functional Grade Functional Grade
          • Unconjugated
          • eFluor 450
          • PE 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          500 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2
          Contains
          no preservative
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_469213
          Product Specific Information
          Description: The TRFK5 antibody reacts with mouse and human interleukin-5 (IL-5). The TRFK5 antibody is a neutralizing antibody. Mouse IL-5 is a disulfide-linked homodimer, containing two 113 amino acid peptides; these resolve as 32-34 kDa bands in SDS-PAGE. IL-5 is produced by T cells and has been known as eosinophil-differentiating factor (EDF), B cell growth factor II (BCGFII), and T cell-replacing factor (TRF). IL-5 induces eosinophil differentiation and promotes eosinophil survival and activation. In mice, IL-5 has been shown to stimulate B cell proliferation and antibody production.
          Applications Reported:The TRFK5 antibody has been reported for use in ELISA capture, ELISPOT capture, neutralization of IL-5 bioactivity, and IHC.
          Applications Tested: The Functional Grade Purified TRFK5 antibody has been tested by LAL assay to verify low endotoxin levels, by ELISA, and in bioassay for neutralization of mouse IL-5 bioactivity.
          The TRFK5 antibody at 0.06 µg/mL has been found to inhibit by 50% the biological effects of 1.0 ng/mL mouse IL-5 in an MC/9 cell proliferation assay. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in the BestProtocols® section.
          The TRFK5 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse IL-5 in combination with the biotin TRFK4 (13-7051) antibody for detection and recombinant mouse IL-5 (14-8051) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 µg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. The TRFK5 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human IL-5 in combination with the biotinylated JES1-5A10 (13-7059) antibody for detection and recombinant human IL-5 (14-80591) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 µg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate.
          Storage and handling: Use in a sterile environment.
          Filtration: 0.2 µm post-manufacturing filtered.
          Purity: Greater than 90%, as determined by SDS-PAGE.
          Endotoxin Level:Less than 0.001 ng/µg antibody, as determined by LAL assay.
          Aggregation:Less than 10%, as determined by HPLC.
          Target Information
          IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.
          Bioinformatics
          Protein Aliases: B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; Cytotoxic T-lymphocyte inducer; Eosinophil differentiation factor; IL-5; ILN; Interleukin; Interleukin-5; Interleukin5; T-cell replacing factor; TRF; unnamed protein product
          View more View less
          Gene Aliases: EDF; IL-5; IL5; TRF
          View more View less
          UniProt ID: (Human) P05113, (Mouse) P04401
          View more View less
          Entrez Gene ID: (Human) 3567, (Mouse) 16191
          View more View less
          Function(s)
          cytokine activity interleukin-5 receptor binding protein binding growth factor activity
          Process(es)
          immune response positive regulation of cell proliferation cytokine-mediated signaling pathway positive regulation of B cell proliferation positive regulation of eosinophil differentiation positive regulation of transcription, DNA-templated positive regulation of JAK-STAT cascade positive regulation of peptidyl-tyrosine phosphorylation positive regulation of immunoglobulin secretion positive regulation of sequence-specific DNA binding transcription factor activity positive regulation of podosome assembly positive regulation of immunoglobulin production inflammatory response eosinophil differentiation interleukin-5-mediated signaling pathway positive regulation of transcription from RNA polymerase II promoter
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline