    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NLGN3 Antibodies
          Rockland
          Neuroligin 3 Monoclonal Antibody (S110-29), FITC
          View all (33) NLGN3 antibodies
          Tech Support
          Cite Neuroligin 3 Monoclonal Antibody (S110-29), FITC
          Additional Information:
          {{banner.description}}
          Neuroligin 3 Monoclonal Antibody (S110-29), FITC
          Product Details
          200-302-W83
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (IHC)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          S110-29
          Immunogen
          Anti-Neuroligin 3 Antibody was produced by repeated immunization of mice with a fusion protein containing amino acids 730-848 of rat Neuroligin-3.
          3D Epitope / Immunogen
          Conjugate
          FITC FITC FITC
          • Unconjugated
          • Biotin 
          • HRP
          • PE 
          • PerCP 
          • Request custom conjugation
          Excitation/Emission Max
          498/517 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.2, with 50% glycerol
          Contains
          0.1% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
          Bioinformatics
          Protein Aliases: ASPGX1; AUTSX1; Gliotactin homolog; neuroligin 3 protein; Neuroligin-3; unnamed protein product
          View more View less
          Gene Aliases: A230085M13Rik; HNL3; KIAA1480; NL3; NLGN3
          View more View less
          UniProt ID: (Human) Q9NZ94, (Mouse) Q8BYM5, (Rat) Q62889
          View more View less
          Entrez Gene ID: (Human) 54413, (Mouse) 245537, (Rat) 171297
          View more View less
          Function(s)
          protein binding signaling receptor activity neurexin family protein binding cell adhesion molecule binding scaffold protein binding receptor activity carboxylic ester hydrolase activity cell adhesion molecule
          Process(es)
          regulation of respiratory gaseous exchange by neurological system process receptor-mediated endocytosis cell adhesion neuron cell-cell adhesion chemical synaptic transmission synapse assembly learning adult behavior social behavior synaptic vesicle endocytosis axon extension modulation of synaptic transmission synapse organization positive regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic rhythmic synaptic transmission inhibitory postsynaptic potential vocalization behavior postsynaptic membrane assembly presynaptic membrane assembly presynapse assembly positive regulation of glutamate receptor signaling pathway positive regulation of excitatory postsynaptic potential visual learning oligodendrocyte differentiation regulation of synaptic transmission, glutamatergic excitatory postsynaptic potential positive regulation of dendritic spine development regulation of dendritic spine morphogenesis positive regulation of inhibitory postsynaptic potential regulation of long-term synaptic potentiation positive regulation of protein localization to synapse regulation of N-methyl-D-aspartate selective glutamate receptor activity regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity regulation of terminal button organization positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity metabolic process
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline