    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • IL-17A Antibodies
          Invitrogen
          IL-17A Polyclonal Antibody, Biotin, PeproTech®
          1 Reference
          View all (187) IL-17A antibodies
          Tech Support
          Cite IL-17A Polyclonal Antibody, Biotin, PeproTech®
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          IL-17A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          IL-17A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          IL-17A Antibody (500-P265BT-50UG) in WB
          Western Blot: To detect mIL-17A by Western Blot analysis IL-17A Polyclonal Antibody, Biotin (Product # 500-P265BT-500UG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mIL-17A is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          IL-17A Antibody in Western Blot (WB)
          IL-17A Antibody in Western Blot (WB)
          IL-17A Antibody in ELISA (ELISA)
          IL-17A Polyclonal Antibody, Biotin, PeproTech®
          Product Details
          500-P265BT-50UG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          0.1-0.2 µg/mL
          -
          ELISA (ELISA)
          0.25-1.0 µg/mL
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Rabbit
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          E.coli-derived Recombinant Murine IL-17A
          3D Epitope / Immunogen
          Conjugate
          Biotin Biotin Biotin
          Form
          Lyophilized
          Concentration
          0.1-1.0 mg/mL
          Amount
          50 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS
          Contains
          no preservative
          Storage conditions
          -20°C
          Shipping conditions
          Ambient
          RRID
          AB_2930151
          Product Specific Information
          AA Sequence of recombinant protein: AAIIPQSSAC PNTEAKDFLQ NVKVNLKVFN SLGAKVSSRR PSDYLNRSTS PWTLHRNEDP DRYPSVIWEA QCRHQRCVNA EGKLDHHMNS VLIQQEILVL KREPESCPFT FRVEKMLVGV GCTCVASIVR QAA.
          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-17A. Anti-Murine IL-17A-specific antibody was purified by affinity chromatography and then biotinylated.
          Sandwich ELISA: To detect mIL-17A by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.25-1.0 µg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine IL-17A (500-P265) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mIL-17A.
          Western Blot: To detect mIL-17A by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mIL-17A is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
          500-P265BT-1MG will be provided as 2 x 500 µg
          Target Information
          Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
          Bioinformatics
          Protein Aliases: CTLA-8; Cytotoxic T-lymphocyte-associated antigen 8; il 17; IL-17; ILN; Interleukin; interleukin 17; Interleukin-17A; Interleukin17A; R-IL-17-A
          View more View less
          Gene Aliases: Ctla-8; Ctla8; IL-17; IL-17A; Il17; Il17a
          View more View less
          UniProt ID: (Mouse) Q62386
          View more View less
          Entrez Gene ID: (Mouse) 16171
          View more View less
          Function(s)
          cytokine activity
          Process(es)
          inflammatory response positive regulation of necrotic cell death positive regulation of interleukin-23 production positive regulation of osteoclast differentiation positive regulation of transcription from RNA polymerase II promoter cellular response to interleukin-1 cellular response to glucocorticoid stimulus fibroblast activation positive regulation of cytokine production involved in inflammatory response
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline