    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Arp3 Antibodies
          Proteintech
          ARP3/ARP3B Monoclonal Antibody (1D12C7)
          View all (18) Arp3 antibodies
          Tech Support
          Cite ARP3/ARP3B Monoclonal Antibody (1D12C7)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          ARP3/ARP3B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ARP3/ARP3B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          ARP3/ARP3B Antibody (68439-1-IG) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed HUVEC cells using ARP3/ARP3B antibody (68439-1-Ig, Clone: 1D12C7) at dilution of 1:800 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ARP3/ARP3B Antibody in Immunocytochemistry (ICC/IF)
          ARP3/ARP3B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ARP3/ARP3B Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ARP3/ARP3B Antibody in Western Blot (WB)
          ARP3/ARP3B Monoclonal Antibody (1D12C7)
          Product Details
          68439-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:500-1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:400-1:1,600
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rabbit, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1D12C7
          Immunogen
          ARP3/ARP3B fusion protein Ag5017
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          150 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: RQVGERTLTG TVIDSGDGVT HVIPVAEGYV IGSCIKHIPI AGRDITYFIQ QLLRDREVGI PPEQSLETAK AVKERYSYVC PDLVKEFNKY DTDGSKWIKQ YTGINAISKK EFSIDVGYER FLGPEIFFHP EFANPDFTQP ISEVVDEVIQ NCPIDVRRPL YKNIVLSGGS TMFRDFGRRL QRDLKRTVDA RLKLSEELSG GRLKPKPIDV QVITHHMQRY AVWFGGSMLA STPEFYQVCH TKKDYEEIGP SICRHNPVFG VMS
          Target Information
          The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.
          Bioinformatics
          Protein Aliases: Actin-like protein 3; Actin-related protein 3; actin-related protein 3 homolog (yest); ARP3 actin related protein 3 homolog; ARP3 actin-related protein 3 homolog; one of seven subunits of the Arp2/3 protein complex; unnamed protein product
          View more View less
          Gene Aliases: 1200003A09Rik; ACTR3; ARP3
          View more View less
          UniProt ID: (Human) P61158, (Rat) Q4V7C7, (Mouse) Q99JY9
          View more View less
          Entrez Gene ID: (Rabbit) 100008790, (Human) 10096, (Pig) 396738, (Rat) 81732, (Mouse) 74117
          View more View less
          Function(s)
          nucleotide binding actin binding structural constituent of cytoskeleton protein binding ATP binding actin filament binding actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          establishment or maintenance of cell polarity asymmetric cell division positive regulation of lamellipodium assembly meiotic chromosome movement towards spindle pole cell projection organization meiotic cytokinesis Arp2/3 complex-mediated actin nucleation positive regulation of transcription from RNA polymerase II promoter meiotic cell cycle spindle localization cilium morphogenesis actin polymerization-dependent cell motility cellular response to interferon-gamma response to carbohydrate positive regulation of actin filament polymerization regulation of myosin II filament organization positive regulation of neuron differentiation response to antibiotic positive regulation of dendrite morphogenesis positive regulation of filopodium assembly actin filament organization
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline