Sequence of this protein is as follows: RQVGERTLTG TVIDSGDGVT HVIPVAEGYV IGSCIKHIPI AGRDITYFIQ QLLRDREVGI PPEQSLETAK AVKERYSYVC PDLVKEFNKY DTDGSKWIKQ YTGINAISKK EFSIDVGYER FLGPEIFFHP EFANPDFTQP ISEVVDEVIQ NCPIDVRRPL YKNIVLSGGS TMFRDFGRRL QRDLKRTVDA RLKLSEELSG GRLKPKPIDV QVITHHMQRY AVWFGGSMLA STPEFYQVCH TKKDYEEIGP SICRHNPVFG VMS
The specific function of this gene has not yet been determined; however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion.
Protein Aliases: Actin-like protein 3; Actin-related protein 3; actin-related protein 3 homolog (yest); ARP3 actin related protein 3 homolog; ARP3 actin-related protein 3 homolog; one of seven subunits of the Arp2/3 protein complex; unnamed protein product
Gene Aliases: 1200003A09Rik; ACTR3; ARP3
UniProt ID: (Human) P61158, (Rat) Q4V7C7, (Mouse) Q99JY9
Entrez Gene ID: (Rabbit) 100008790, (Human) 10096, (Pig) 396738, (Rat) 81732, (Mouse) 74117