    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • OTUD6B Antibodies
          Proteintech
          OTUD6B Monoclonal Antibody (1D1G1)
          View all (9) OTUD6B antibodies
          Tech Support
          Cite OTUD6B Monoclonal Antibody (1D1G1)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          OTUD6B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          OTUD6B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          OTUD6B Antibody (68450-1-IG) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed PC-3 cells using OTUD6B antibody (68450-1-Ig, Clone: 1D1G1) at dilution of 1:800 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L) (SA00013-1). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          OTUD6B Antibody in Immunocytochemistry (ICC/IF)
          OTUD6B Antibody in Western Blot (WB)
          OTUD6B Monoclonal Antibody (1D1G1)
          Product Details
          68450-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunocytochemistry (ICC/IF)
          1:800
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1D1G1
          Immunogen
          OTUD6B fusion protein Ag33607
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MEAVLTEELD EEEQLLRRHR KEKKELQAKI QGMKNAVPKN DKKRRKQLTE DVAKLEKEME QKHREELEQL KLTTKENKID SVAVNISNLV LENQPPRISK AQKRREKKAA LEKEREERIA EAEIENLTGA RHMESEKLAQ ILAARQLEIK QIPSDGH
          Target Information
          Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA5 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
          Bioinformatics
          Protein Aliases: Deubiquitinase OTUD6B; DUBA-5; ortholog of human CGI-77; OTU domain containing 6B; OTU domain-containing protein 6B; unnamed protein product
          View more View less
          Gene Aliases: 2600013N14Rik; AU015433; CGI-77; DUBA-5; DUBA5; IDDFSDA; OTUD6B; RGD1310024
          View more View less
          UniProt ID: (Human) Q8N6M0, (Mouse) Q8K2H2
          View more View less
          Entrez Gene ID: (Human) 51633, (Rat) 297911, (Mouse) 72201
          View more View less
          Function(s)
          thiol-dependent ubiquitin-specific protease activity protein binding peptidase activity cysteine-type peptidase activity hydrolase activity molecular_function cysteine protease
          Process(es)
          proteolysis cell proliferation protein deubiquitination negative regulation of translation proteasome assembly positive regulation of translation biological_process
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0