Sequence of this protein is as follows: MEAVLTEELD EEEQLLRRHR KEKKELQAKI QGMKNAVPKN DKKRRKQLTE DVAKLEKEME QKHREELEQL KLTTKENKID SVAVNISNLV LENQPPRISK AQKRREKKAA LEKEREERIA EAEIENLTGA RHMESEKLAQ ILAARQLEIK QIPSDGH
Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA5 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
Protein Aliases: Deubiquitinase OTUD6B; DUBA-5; ortholog of human CGI-77; OTU domain containing 6B; OTU domain-containing protein 6B; unnamed protein product
Gene Aliases: 2600013N14Rik; AU015433; CGI-77; DUBA-5; DUBA5; IDDFSDA; OTUD6B; RGD1310024
UniProt ID: (Human) Q8N6M0, (Mouse) Q8K2H2
Entrez Gene ID: (Human) 51633, (Rat) 297911, (Mouse) 72201