    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • GIMA7 Antibodies
          Proteintech
          GIMAP7 Monoclonal Antibody (3E4A6)
          View all (5) GIMA7 antibodies
          Tech Support
          Cite GIMAP7 Monoclonal Antibody (3E4A6)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          GIMAP7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GIMAP7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GIMAP7 Antibody (68451-1-IG) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using GIMAP7 antibody (68451-1-Ig, Clone: 3E4A6) at dilution of 1:800 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GIMAP7 Antibody in Immunocytochemistry (ICC/IF)
          GIMAP7 Antibody in Western Blot (WB)
          GIMAP7 Antibody in Immunoprecipitation (IP)
          GIMAP7 Monoclonal Antibody (3E4A6)
          Product Details
          68451-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:50,000
          -
          Immunocytochemistry (ICC/IF)
          1:400-1:1,600
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          0.5-4.0 µg for 1.0-3.0 mg of total protein lysate
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3E4A6
          Immunogen
          GIMAP7 fusion protein Ag10740
          View immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          150 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAESEDRSLR IVLVGKTGSG KSATANTILG EEIFDSRIAA QAVTKNCQKA SREWQGRDLL VVDTPGLFDT KESLDTTCKE ISRCIISSCP GPHAIVLVLL LGRYTEEEQK TVALIKAVFG KSAMKHMVIL FTRKEELEGQ SFHDFIADAD VGLKSIVKEC GNRCCAFSNS KKTSKAEKES QVQELVELIE KMVQCNEGAY FSDDIYKDTE ERLKQREEVL RKIYTDQLNE EIKLVEEDKH KSEEEKEKEI KLLKLKYDEK IKNIREEAER NIFKDVFNRI WKMLSEIWHR FLSKCKFYSS
          Target Information
          GIMAP7 is also name as hIAN7 or IAN 7. The function of GIMAP7 remains largely unknown.
          Bioinformatics
          Protein Aliases: GTPase IMAP family member 7; IAN-7; immune associated nucleotide; immune-associated nucleotide-binding protein 7; Immunity-associated nucleotide 7 protein
          View more View less
          Gene Aliases: GIMAP7; hIAN7; IAN7
          View more View less
          UniProt ID: (Human) Q8NHV1
          View more View less
          Entrez Gene ID: (Human) 168537
          View more View less
          Function(s)
          nucleotide binding GTPase activity protein binding GTP binding identical protein binding protein homodimerization activity
          Process(es)
          GTP metabolic process
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline