    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • GIMA7 Antibodies
          Proteintech
          GIMAP7 Monoclonal Antibody (3E4A6), CoraLite® Plus 488
          View all (6) GIMA7 antibodies
          Tech Support
          Cite GIMAP7 Monoclonal Antibody (3E4A6), CoraLite® Plus 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          GIMAP7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GIMAP7 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          GIMAP7 Antibody (CL488-68451) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite® Plus 488 GIMAP7 antibody (CL488-68451, Clone: 3E4A6) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GIMAP7 Antibody in Immunocytochemistry (ICC/IF)
          GIMAP7 Monoclonal Antibody (3E4A6), CoraLite® Plus 488
          Product Details
          CL488-68451
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3E4A6
          Immunogen
          GIMAP7 fusion protein Ag10740
          View immunogen
          Conjugate
          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          493/522 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAESEDRSLR IVLVGKTGSG KSATANTILG EEIFDSRIAA QAVTKNCQKA SREWQGRDLL VVDTPGLFDT KESLDTTCKE ISRCIISSCP GPHAIVLVLL LGRYTEEEQK TVALIKAVFG KSAMKHMVIL FTRKEELEGQ SFHDFIADAD VGLKSIVKEC GNRCCAFSNS KKTSKAEKES QVQELVELIE KMVQCNEGAY FSDDIYKDTE ERLKQREEVL RKIYTDQLNE EIKLVEEDKH KSEEEKEKEI KLLKLKYDEK IKNIREEAER NIFKDVFNRI WKMLSEIWHR FLSKCKFYSS
          Target Information
          GIMAP7 is also name as hIAN7 or IAN 7. The function of GIMAP7 remains largely unknown.
          Bioinformatics
          Protein Aliases: GTPase IMAP family member 7; IAN-7; immune associated nucleotide; immune-associated nucleotide-binding protein 7; Immunity-associated nucleotide 7 protein
          View more View less
          Gene Aliases: GIMAP7; hIAN7; IAN7
          View more View less
          UniProt ID: (Human) Q8NHV1
          View more View less
          Entrez Gene ID: (Human) 168537
          View more View less
          Function(s)
          nucleotide binding GTPase activity protein binding GTP binding identical protein binding protein homodimerization activity
          Process(es)
          GTP metabolic process
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline