    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • UNC13A Antibodies
          Proteintech
          UNC13A Monoclonal Antibody (3C3A9)
          Tech Support
          Cite UNC13A Monoclonal Antibody (3C3A9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          UNC13A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UNC13A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          UNC13A Antibody (68483-1-IG) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed SH-SY5Y cells using UNC13A antibody (68483-1-Ig, Clone: 3C3A9) at dilution of 1:800 and CoraLite®488-Conjugated Goat Anti-Mouse IgG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          UNC13A Antibody in Immunocytochemistry (ICC/IF)
          UNC13A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UNC13A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          UNC13A Antibody in Western Blot (WB)
          Product Details
          68483-1-IG
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:10,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200-1:1,000
          -
          Immunocytochemistry (ICC/IF)
          1:400-1:1,600
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rabbit, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3C3A9
          Immunogen
          Peptide
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • CoraLite 555
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          150 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Wet ice
          Target Information
          UNC13A, also named as KIAA1032, belongs to the unc-13 family. It plays a role in vesicle maturation during exocytosis as a target of the diacylglycerol second messenger pathway. UNC13A is involved in neurotransmitter release by acting in synaptic vesicle priming prior to vesicle fusion and participates in the activity-dependent refilling of readily releasable vesicle pool (RRP). It is essential for synaptic vesicle maturation in most excitatory/glutamatergic but not inhibitory/GABA-mediated synapses. UNC13A is also involved in secretory granule priming in insulin secretion.
          Bioinformatics
          Protein Aliases: Munc13-1; Protein unc-13 homolog A; unc13a
          View more View less
          Gene Aliases: 2410078G03Rik; KIAA1032; Munc13-1; UNC13A; Unc13h1
          View more View less
          UniProt ID: (Human) Q9UPW8, (Mouse) Q4KUS2, (Rat) Q62768
          View more View less
          Entrez Gene ID: (Human) 23025, (Pig) 100627038, (Mouse) 382018, (Rat) 64829
          View more View less
          Function(s)
          calcium ion binding calmodulin binding phospholipid binding zinc ion binding syntaxin-1 binding diacylglycerol binding metal ion binding SNARE binding protein binding protein domain specific binding protein homodimerization activity protein heterodimerization activity protein N-terminus binding
          Process(es)
          positive regulation of neurotransmitter secretion exocytosis chemical synaptic transmission neurotransmitter secretion neuromuscular junction development regulation of synaptic vesicle priming synaptic vesicle docking synaptic vesicle priming synaptic vesicle maturation synaptic transmission, glutamatergic intracellular signal transduction regulation of short-term neuronal synaptic plasticity beta-amyloid metabolic process long-term synaptic potentiation innervation positive regulation of dendrite extension cell differentiation regulation of synaptic transmission, glutamatergic dense core granule priming neuronal dense core vesicle exocytosis
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline