    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • UNC13A Antibodies
          Proteintech
          UNC13A Monoclonal Antibody (3C3A9), CoraLite® 555
          View all (8) UNC13A antibodies
          Tech Support
          Cite UNC13A Monoclonal Antibody (3C3A9), CoraLite® 555
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          UNC13A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UNC13A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          UNC13A Antibody (CL555-68483) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed SH-SY5Y cells using CoraLite®555 UNC13A antibody (CL555-68483, Clone: 3C3A9) at dilution of 1:400. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          UNC13A Antibody in Immunocytochemistry (ICC/IF)
          UNC13A Antibody in Flow Cytometry (Flow)
          Product Details
          CL555-68483
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rabbit, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3C3A9
          Immunogen
          Peptide
          Conjugate
          CoraLite® 555 CoraLite® 555 CoraLite® 555
          • Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          555/570 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          UNC13A, also named as KIAA1032, belongs to the unc-13 family. It plays a role in vesicle maturation during exocytosis as a target of the diacylglycerol second messenger pathway. UNC13A is involved in neurotransmitter release by acting in synaptic vesicle priming prior to vesicle fusion and participates in the activity-dependent refilling of readily releasable vesicle pool (RRP). It is essential for synaptic vesicle maturation in most excitatory/glutamatergic but not inhibitory/GABA-mediated synapses. UNC13A is also involved in secretory granule priming in insulin secretion.
          Bioinformatics
          Protein Aliases: Munc13-1; Protein unc-13 homolog A; unc13a
          View more View less
          Gene Aliases: 2410078G03Rik; KIAA1032; Munc13-1; UNC13A; Unc13h1
          View more View less
          UniProt ID: (Human) Q9UPW8, (Mouse) Q4KUS2, (Rat) Q62768
          View more View less
          Entrez Gene ID: (Human) 23025, (Pig) 100627038, (Mouse) 382018, (Rat) 64829
          View more View less
          Function(s)
          calcium ion binding calmodulin binding phospholipid binding zinc ion binding syntaxin-1 binding diacylglycerol binding metal ion binding SNARE binding protein binding protein domain specific binding protein homodimerization activity protein heterodimerization activity protein N-terminus binding
          Process(es)
          positive regulation of neurotransmitter secretion exocytosis chemical synaptic transmission neurotransmitter secretion neuromuscular junction development regulation of synaptic vesicle priming synaptic vesicle docking synaptic vesicle priming synaptic vesicle maturation synaptic transmission, glutamatergic intracellular signal transduction regulation of short-term neuronal synaptic plasticity beta-amyloid metabolic process long-term synaptic potentiation innervation positive regulation of dendrite extension cell differentiation regulation of synaptic transmission, glutamatergic dense core granule priming neuronal dense core vesicle exocytosis
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline