    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • IP3 Receptor 1 Antibodies
          Antibodies Inc
          IP3 Receptor/Type 1 Monoclonal Antibody (L24/18), FL594
          View all (29) IP3 Receptor 1 antibodies
          Tech Support
          Cite IP3 Receptor/Type 1 Monoclonal Antibody (L24/18), FL594
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          IP3 Receptor/Type 1 Monoclonal Antibody (L24/18), FL594
          Product Details
          75-035-FL594
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Dog, Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          L24/18
          Immunogen
          Fusion protein amino acids 2680-2749 (cytoplasmic carboxyl terminus) of rat type 1 IP3R (accession number P29994) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Intracellular channel that mediates calcium release from the endoplasmic reticulum following stimulation by inositol 1,4,5-trisphosphate. Involved in the regulation of epithelial secretion of electrolytes and fluid through the interaction with AHCYL1. Plays a role in ER stress-induced apoptosis. Cytoplasmic calcium released from the ER triggers apoptosis by the activation of CaM kinase II, eventually leading to the activation of downstream apoptosis pathways.
          Bioinformatics
          Protein Aliases: inositol 1,4,5 trisphosphate receptor; inositol 1,4,5-triphosphate receptor 1; inositol 1,4,5-triphosphate receptor, type 1; inositol 1,4,5-trisphosphate receptor type 1; inositol 1,4,5-trisphosphate-binding protein P400; inositol-1,4,5-triphosphate receptor; inositol-1,4,5-triphosphate receptor (alt., clone pI16); InsP>3
          View more View less
          Gene Aliases: ACV; CLA4; D6Pas2; Gm10429; I145TR; InsP3R; INSP3R1; IP3R; IP3R 1; IP3R1; Itpr-1; opt; P400; Pcp-1; Pcp1; PPP1R94; SCA15; SCA16; SCA29; wblo
          View more View less
          UniProt ID: (Human) Q14643, (Mouse) P11881, (Rat) P29994
          View more View less
          Entrez Gene ID: (Human) 3708, (Dog) 476548, (Mouse) 16438, (Rat) 25262
          View more View less
          Function(s)
          ion channel activity inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity calcium channel activity calcium ion binding protein binding calcium ion transmembrane transporter activity calcium-release channel activity calcium channel inhibitor activity protein domain specific binding phosphatidylinositol binding protein homodimerization activity inositol 1,4,5 trisphosphate binding inositol 1,4,5-trisphosphate receptor activity involved in regulation of postsynaptic cytosolic calcium levels ATP binding protein phosphatase binding identical protein binding ion channel binding macromolecular complex binding ligand-gated ion channel
          Process(es)
          cell morphogenesis response to hypoxia ion transport calcium ion transport signal transduction phospholipase C-activating G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled acetylcholine receptor signaling pathway single fertilization regulation of autophagy calcium-mediated signaling calcium ion transmembrane import into mitochondrion epithelial fluid transport positive regulation of apoptotic process regulation of calcium-mediated signaling negative regulation of calcium-mediated signaling release of sequestered calcium ion into cytosol protein homotetramerization regulation of cytosolic calcium ion concentration transmembrane transport intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress calcium ion transmembrane transport regulation of postsynaptic cytosolic calcium ion concentration release of sequestered calcium ion into cytosol by endoplasmic reticulum positive regulation of cytosolic calcium ion concentration post-embryonic development positive regulation of neuron projection development endoplasmic reticulum calcium ion homeostasis negative regulation of apoptotic process voluntary musculoskeletal movement positive regulation of calcium ion transport cellular response to cAMP liver regeneration positive regulation of hepatocyte proliferation dendrite development cellular response to hypoxia
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-5bc99cc877-v966v:80/100.66.72.244:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0