    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CACNB1 Antibodies
          Antibodies Inc
          Cavbeta1 Ca2+ Channel Monoclonal Antibody (N7/18)
          View all (17) CACNB1 antibodies
          Tech Support
          Cite Cavbeta1 Ca2+ Channel Monoclonal Antibody (N7/18)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          Cavbeta1 Ca2+ Channel Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cavbeta1 Ca2+ Channel Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          Cavbeta1 Ca2+ Channel Antibody (75-052) in ICC/IF
          When coexpressed with CaV1.1 in F8 (CaV1.1-/-, STAC3-/-) myotubes, STAC3-ETLAAA-GFP does not colocalize in clusters with the β1 subunit, contrary to wild-type STAC3-GFP. Image from publication CC-BY-4.0. PMID:36161458. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Cavbeta1 Ca2+ Channel Antibody in Immunocytochemistry (ICC/IF)
          Cavbeta1 Ca2+ Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cavbeta1 Ca2+ Channel Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cavbeta1 Ca2+ Channel Antibody in Western Blot (WB)
          Cavbeta1 Ca2+ Channel Antibody in Western Blot (WB)
          Cavbeta1 Ca2+ Channel Monoclonal Antibody (N7/18)
          Product Details
          75-052
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          1:250
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          2 µg
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Non-human primate, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N7/18
          Immunogen
          Synthetic peptide amino acids 19-34 of rat Cav1 (PMEVFDPSPQGKYSKR; accession number NP_059042).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Calcium Channel 1 is a protein encoded by the CACNB1 gene and belongs to the calcium channel beta subunit family. Calcium Channel 1 plays an important role in the calcium channel by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Calcium Channel 1 is involved in modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Diseases associated with Calcium Channel 1 protein dysfunction include malignant hyperthermia susceptibility.
          Bioinformatics
          Protein Aliases: brain calcium channel beta 1 subunit; CAB1; calcium channel beta 1 subunit; Calcium channel voltage-dependent subunit beta 1; calcium channel, L type, beta 1 polypeptide; calcium channel, voltage-dependent, beta 1 subunit; dihydropyridine-sensitive L-type, calcium channel beta-1 subunit; MGC41896; unnamed protein product; voltage-dependent calcium channel beta1 subunit; Voltage-dependent L-type calcium channel subunit beta-1
          View more View less
          Gene Aliases: CAB1; CACNB1; CACNLB1; Cchb1; CCHLB1
          View more View less
          UniProt ID: (Human) Q02641, (Mouse) Q8R3Z5, (Rat) P54283
          View more View less
          Entrez Gene ID: (Human) 782, (Mouse) 12295, (Rat) 50688
          View more View less
          Function(s)
          voltage-gated calcium channel activity calcium channel regulator activity calcium channel activity protein binding high voltage-gated calcium channel activity voltage-gated ion channel activity protein kinase binding protein domain specific binding phosphoprotein binding voltage-gated ion channel
          Process(es)
          ion transport calcium ion transport chemical synaptic transmission ion transmembrane transport positive regulation of muscle contraction calcium ion transmembrane transport regulation of calcium ion transmembrane transport via high voltage-gated calcium channel cellular response to beta-amyloid protein targeting to membrane transport neuromuscular junction development regulation of ion transmembrane transport regulation of voltage-gated calcium channel activity
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline