    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • Calcium Channel beta-4 Antibodies
          Antibodies Inc
          Cavbeta4 Ca2+ Channel Monoclonal Antibody (N10/7), FL650
          View all (17) Calcium Channel beta-4 antibodies
          Tech Support
          Cite Cavbeta4 Ca2+ Channel Monoclonal Antibody (N10/7), FL650
          Additional Information:
          {{banner.description}}
          Cavbeta4 Ca2+ Channel Monoclonal Antibody (N10/7), FL650
          Product Details
          75-054-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          1:250
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N10/7
          Immunogen
          Synthetic peptide amino acids 458-474 of rat Cav4 (DENYHNERARKSRNRLS; accession number D4A055).
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy and juvenile myoclonic epilepsy. Multiple transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: CAB4; calcium channel beta 4 subunit; Calcium channel voltage-dependent subunit beta 4; dihydropyridine-sensitive L-type, calcium channel beta-4 subunit; modulates calcium channel function; OTTHUMP00000207247; OTTHUMP00000207249; unnamed protein product; voltage-dependent calcium channel beta 4 subunit; Voltage-dependent L-type calcium channel subunit beta-4
          View more View less
          Gene Aliases: 3110038O15Rik; CAB4; CACNB4; CACNLB4; Cchb4; EA5; EIG9; EJM; EJM4; EJM6; lh
          View more View less
          UniProt ID: (Human) O00305, (Rat) D4A055, (Mouse) Q8R0S4
          View more View less
          Entrez Gene ID: (Pig) 396585, (Human) 785, (Rat) 58942, (Mouse) 12298
          View more View less
          Function(s)
          voltage-gated calcium channel activity calcium channel regulator activity calcium channel activity protein binding protein kinase binding voltage-gated calcium channel activity involved in regulation of presynaptic cytosolic calcium levels high voltage-gated calcium channel activity voltage-gated ion channel activity voltage-gated ion channel
          Process(es)
          ion transport calcium ion transport gamma-aminobutyric acid signaling pathway chemical synaptic transmission neuromuscular junction development adult walking behavior negative regulation of cell proliferation gamma-aminobutyric acid secretion neuronal action potential propagation ion transmembrane transport synaptic transmission, glutamatergic regulation of membrane potential cAMP metabolic process spleen development thymus development Peyer's patch development T cell receptor signaling pathway neurological system process detection of light stimulus involved in visual perception muscle cell development calcium ion transmembrane transport regulation of presynaptic cytosolic calcium ion concentration positive regulation of protein localization to nucleolus cellular response to leukemia inhibitory factor negative regulation of G1/S transition of mitotic cell cycle regulation of synaptic vesicle exocytosis cellular calcium ion homeostasis T cell differentiation regulation of ion transmembrane transport muscle fiber development synaptic transmission, GABAergic regulation of voltage-gated calcium channel activity transport
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline