    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • Slo3 Antibodies
          Antibodies Inc
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Monoclonal Antibody (N2/16)
          View all (7) Slo3 antibodies
          Tech Support
          Cite KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Monoclonal Antibody (N2/16)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Antibody (75-056) in ICC/IF
          Mouse sperm immunofluorescence staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Antibody in Immunocytochemistry (ICC/IF)
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Antibody in Western Blot (WB)
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Monoclonal Antibody (N2/16)
          Product Details
          75-056
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Xenopus
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N2/16
          Immunogen
          Fusion protein amino acids 1052-1121 of mouse Slo3 (accession number O54982) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The Slo3 channel is a novel potassium channel abundantly expressed in mammalian speramtocytes- tests have shown that it is expressed in both mouse and human testis. It represents a new and unique type of potassium channel that is regulated by both intracellular pH and membrane voltage. Because of its sensitivity to both pH and voltage, Slo3 may play a role in alkalization-mediated K(+) fluxes associated with sperm capacitation.
          Bioinformatics
          Protein Aliases: Calcium-activated potassium channel subunit alpha-3; Calcium-activated potassium channel, subfamily M subunit alpha-3; KSper; mSlo3; pH-sensitive maxi potassium channel; Pore-forming subunit of the sperm-specific alkalization activated K(+) current; Potassium channel subfamily U member 1; potassium large conductance pH-sensitive channel, subfamily M, alpha member 3; Slowpoke homolog 3
          View more View less
          Gene Aliases: Kcnma3; Kcnu1; Ksper; mSlo3; Slo3
          View more View less
          UniProt ID: (Mouse) O54982
          View more View less
          Entrez Gene ID: (Mouse) 16532
          View more View less
          Function(s)
          ion channel activity voltage-gated ion channel activity voltage-gated potassium channel activity potassium channel activity calcium-activated potassium channel activity large conductance calcium-activated potassium channel activity ion channel
          Process(es)
          transport ion transport potassium ion transport regulation of ion transmembrane transport transmembrane transport potassium ion transmembrane transport
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline