    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Slo3 Antibodies
          Antibodies Inc
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Monoclonal Antibody (N2/16), FL594
          View all (7) Slo3 antibodies
          Tech Support
          Cite KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Monoclonal Antibody (N2/16), FL594
          Additional Information:
          {{banner.description}}
          KCNU1/Slo3 pH-Sensitive Maxi-K+ Channel Monoclonal Antibody (N2/16), FL594
          Product Details
          75-056-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Xenopus
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N2/16
          Immunogen
          Fusion protein amino acids 1052-1121 of mouse Slo3 (accession number O54982) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The Slo3 channel is a novel potassium channel abundantly expressed in mammalian speramtocytes- tests have shown that it is expressed in both mouse and human testis. It represents a new and unique type of potassium channel that is regulated by both intracellular pH and membrane voltage. Because of its sensitivity to both pH and voltage, Slo3 may play a role in alkalization-mediated K(+) fluxes associated with sperm capacitation.
          Bioinformatics
          Protein Aliases: Calcium-activated potassium channel subunit alpha-3; Calcium-activated potassium channel, subfamily M subunit alpha-3; KSper; mSlo3; pH-sensitive maxi potassium channel; Pore-forming subunit of the sperm-specific alkalization activated K(+) current; Potassium channel subfamily U member 1; potassium large conductance pH-sensitive channel, subfamily M, alpha member 3; Slowpoke homolog 3
          View more View less
          Gene Aliases: Kcnma3; Kcnu1; Ksper; mSlo3; Slo3
          View more View less
          UniProt ID: (Mouse) O54982
          View more View less
          Entrez Gene ID: (Mouse) 16532
          View more View less
          Function(s)
          ion channel activity voltage-gated ion channel activity voltage-gated potassium channel activity potassium channel activity calcium-activated potassium channel activity large conductance calcium-activated potassium channel activity ion channel
          Process(es)
          transport ion transport potassium ion transport regulation of ion transmembrane transport transmembrane transport potassium ion transmembrane transport
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline