    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • LRRC4 Antibodies
          Antibodies Inc
          NGL-2/LRRC4 Monoclonal Antibody (N50/36)
          View all (15) LRRC4 antibodies
          Tech Support
          Cite NGL-2/LRRC4 Monoclonal Antibody (N50/36)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          NGL-2/LRRC4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NGL-2/LRRC4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          NGL-2/LRRC4 Antibody (75-084) in WB
          Adult rat brain membrane (RBM) and transfected cell immunoblot: extracts of RBM and COS cells transiently transfected with Myc-tagged NGL-1, NGL-2, NGL-3 or untagged Kv2.1 plasmid and probed with N50/36 (left) or K89/41 (right) TC supe. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NGL-2/LRRC4 Antibody in Western Blot (WB)
          NGL-2/LRRC4 Monoclonal Antibody (N50/36)
          Product Details
          75-084
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N50/36
          Immunogen
          Fusion protein amino acids 550-652 of mouse NGL-2 (accession number AAG60620) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The LRRC4 gene, also known as netrin-G ligand 2 (NGL-2), is a brain-specific member of the leucine-rich repeat superfamily, playing a critical role in the central nervous system. Its expression is particularly high in areas such as the hippocampal CA1 and CA2 regions, as well as in granule cells of the dentate gyrus. LRRC4 is a known tumor suppressor gene associated with glioma suppression, as it regulates glioma cell growth, angiogenesis, and invasion through extensive signaling networks. It can participate in multiphase loops involving miRNA, transcription factors, and gene methylation modifications. This gene targets various genes involved in cell motility, impacting glioblastoma cell movement and polarization by binding to enhancer elements and activating transcription, notably the RAP1GAP gene. Its function and expression are crucial in maintaining normal brain function and hold potential implications in the management of gliomagenesis.
          Bioinformatics
          Protein Aliases: Brain tumor-associated protein MBAG1; leucine rich repeat containing 4 protein; Leucine-rich repeat-containing protein 4; LIBG-like protein; LRRC4-like protein; Netrin-G2 ligand; NGL-2; NLG2; protein NGL-2
          View more View less
          Gene Aliases: Lrrc4; MBAG1; Nag14; NGL-2; NLG-2
          View more View less
          UniProt ID: (Mouse) Q99PH1, (Rat) Q45R42
          View more View less
          Entrez Gene ID: (Mouse) 192198, (Rat) 641521
          View more View less
          Function(s)
          protein binding
          Process(es)
          regulation of synapse organization synapse organization postsynaptic density protein 95 clustering
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline