    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • LRRC4 Antibodies
          Antibodies Inc
          NGL-2/LRRC4 Monoclonal Antibody (N50/36), FL490
          View all (15) LRRC4 antibodies
          Tech Support
          Cite NGL-2/LRRC4 Monoclonal Antibody (N50/36), FL490
          Additional Information:
          {{banner.description}}
          NGL-2/LRRC4 Monoclonal Antibody (N50/36), FL490
          Product Details
          75-084-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N50/36
          Immunogen
          Fusion protein amino acids 550-652 of mouse NGL-2 (accession number AAG60620) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          The LRRC4 gene, also known as netrin-G ligand 2 (NGL-2), is a brain-specific member of the leucine-rich repeat superfamily, playing a critical role in the central nervous system. Its expression is particularly high in areas such as the hippocampal CA1 and CA2 regions, as well as in granule cells of the dentate gyrus. LRRC4 is a known tumor suppressor gene associated with glioma suppression, as it regulates glioma cell growth, angiogenesis, and invasion through extensive signaling networks. It can participate in multiphase loops involving miRNA, transcription factors, and gene methylation modifications. This gene targets various genes involved in cell motility, impacting glioblastoma cell movement and polarization by binding to enhancer elements and activating transcription, notably the RAP1GAP gene. Its function and expression are crucial in maintaining normal brain function and hold potential implications in the management of gliomagenesis.
          Bioinformatics
          Protein Aliases: Brain tumor-associated protein MBAG1; leucine rich repeat containing 4 protein; Leucine-rich repeat-containing protein 4; LIBG-like protein; LRRC4-like protein; Netrin-G2 ligand; NGL-2; NLG2; protein NGL-2
          View more View less
          Gene Aliases: Lrrc4; MBAG1; Nag14; NGL-2; NLG-2
          View more View less
          UniProt ID: (Mouse) Q99PH1, (Rat) Q45R42
          View more View less
          Entrez Gene ID: (Mouse) 192198, (Rat) 641521
          View more View less
          Function(s)
          protein binding
          Process(es)
          regulation of synapse organization synapse organization postsynaptic density protein 95 clustering
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline