    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • STRN4 Antibodies
          Antibodies Inc
          Zinedin Monoclonal Antibody (K88/64), FL490
          View all (16) STRN4 antibodies
          Questions & Answers
          Cite Zinedin Monoclonal Antibody (K88/64), FL490
          Zinedin Monoclonal Antibody (K88/64), FL490
          Product Details
          75-099-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          K88/64
          Immunogen
          Fusion protein amino acids 42-354 of human Zinedin/Striatin-4 (accession number Q9NRL3) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Binds calmodulin in a calcium dependent manner. May function as scaffolding or signaling protein.
          Bioinformatics
          Protein Aliases: protein phosphatase 2 regulatory subunit B'''gamma; striatin, calmodulin binding protein 4; Striatin-4; Zinedin
          View more View less
          Gene Aliases: PPP2R6C; STRN4; ZIN; zinedin
          View more View less
          UniProt ID: (Human) Q9NRL3
          View more View less
          Entrez Gene ID: (Human) 29888, (Rat) 308392
          View more View less
          Function(s)
          protein binding calmodulin binding protein complex binding protein phosphatase 2A binding armadillo repeat domain binding protein domain specific binding
          Process(es)
          biological_process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-55ff9ddd88-xq6xf:80/100.66.76.183:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0