    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • QKI Antibodies
          Antibodies Inc
          Pan-QKI Monoclonal Antibody (N147/6)
          View all (41) QKI antibodies
          Tech Support
          Cite Pan-QKI Monoclonal Antibody (N147/6)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          Pan-QKI Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          Pan-QKI Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 7
          Pan-QKI Antibody (75-168) in IHC (F)
          Representative confocal images of colocalisation of EdU+ and Pan-QKI in the dorsal WM of mouse spinal cord. Closed arrows denote colocalised cells, open arrows denote non-colocalised cells. Image from publication CC-BY-4.0. PMID:36336067. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Pan-QKI Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Pan-QKI Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Pan-QKI Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Pan-QKI Antibody in Western Blot (WB)
          Pan-QKI Antibody in Western Blot (WB)
          Pan-QKI Antibody in Western Blot (WB)
          Pan-QKI Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Pan-QKI Monoclonal Antibody (N147/6)
          Product Details
          75-168
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N147/6
          Immunogen
          Fusion protein amino acids 1-341 (full-length) of human QKI-5 (accession number Q96PU8) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          This gene encodes the sacsin protein, which includes a UbL domain at the N-terminus, a DnaJ domain, and a HEPN domain at the C-terminus. The gene is highly expressed in the central nervous system, also found in skin, skeletal muscles and at low levels in the pancreas. This gene includes a very large exon spanning more than 12.8 kb. Mutations in this gene result in autosomal recessive spastic ataxia of Charlevoix-Saguenay (ARSACS), a neurodegenerative disorder characterized by early-onset cerebellar ataxia with spasticity and peripheral neuropathy. Sacsin has been found to protect against mutant ataxin-1. A pseudogene associated with this gene is located on chromosome 11. Alternative splicing of this gene results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: DKFZp586I0923; homolog of mouse quaking QKI (KH domain RNA binding protein); lethal, Chr 17, U Wisconsin 1; protein quaking; protein quaking-like; QKI; QKI-5 protein; QKI-6 protein; QKI-7 protein; QKI-7b protein; QKI/LOC100132735 fusion; quaking; quaking homolog, KH domain RNA binding; quaking type II; quaking, KH domain containing RNA binding; QUAKING-3; RNA Binding (QKI); RNA binding protein HQK; rqkI; unnamed protein product
          View more View less
          Gene Aliases: 1110003F05Rik; 1500005P18; Hqk; hqkI; l(17)-1Wis; l17Wis1; QK; QK1; QK3
          View more View less
          UniProt ID: (Human) Q96PU8, (Mouse) Q9QYS9
          View more View less
          Entrez Gene ID: (Human) 9444, (Mouse) 19317, (Rat) 499022
          View more View less
          Function(s)
          nucleic acid binding transcription coactivator activity RNA binding mRNA binding mRNA 3'-UTR binding protein binding miRNA binding internal N(7)-methylguanine-containing RNA reader activity
          Process(es)
          vasculogenesis mRNA processing spermatid development intracellular mRNA localization axon ensheathment positive regulation of gene expression negative regulation of cardiac muscle cell apoptotic process regulation of epithelial to mesenchymal transition positive regulation of neuron projection development microglia differentiation negative regulation of angiogenesis negative regulation of translation positive regulation of myelination negative regulation of type I interferon production stress granule assembly vascular smooth muscle cell differentiation myelination muscle cell differentiation long-chain fatty acid biosynthetic process positive regulation of cholesterol biosynthetic process regulation of macrophage differentiation negative regulation of macrophage differentiation positive regulation of transcription, DNA-templated regulation of mRNA splicing, via spliceosome mRNA stabilization regulation of astrocyte differentiation positive regulation of oligodendrocyte differentiation mRNA transport 3'-UTR-mediated mRNA destabilization negative regulation of cold-induced thermogenesis spliceosome-depend formation of circular RNA negative regulation of 3'-UTR-mediated mRNA stabilization positive regulation of 3'-UTR-mediated mRNA stabilization myofibroblast contraction negative regulation of miRNA catabolic process regulation of cell development regulation of multicellular organismal development
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-7fb9f846c9-j9xx9:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0