          • Primary Antibodies ›
          • GRK6 Antibodies
          Antibodies Inc
          GRK6A/B Monoclonal Antibody (N142/28)
          View all (26) GRK6 antibodies
          Tech Support
          Cite GRK6A/B Monoclonal Antibody (N142/28)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          GRK6A/B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GRK6A/B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          GRK6A/B Antibody (75-176) in WB
          Adult rat brain membrane (RBM) and transfected cell immunoblot: extracts of RBM and COS cells transiently transfected with untagged GRK5, GRK6 and Kv2.1 plasmids and probed with N142/28 TC supe. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GRK6A/B Antibody in Western Blot (WB)
          GRK6A/B Monoclonal Antibody (N142/28)
          Product Details
          75-176
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N142/28
          Immunogen
          Fusion protein amino acids 371-576 (C-terminus) of mouse GRK6A (accession number O70293) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          GRK6 is a member of the guanine nucleotide-binding protein coupled receptor kinase subfamily of the serine/threonine protein kinase family. GRK6 phosphorylates the activated forms of G protein-coupled receptors to initiate their deactivation. Several transcript variants encoding different isoforms have been described for GRK6.
          Bioinformatics
          Protein Aliases: EC 2.7.11.16; G protein-coupled receptor kinase 6; G protein-coupled receptor kinase GRK6; kinase GRK6
          View more View less
          Gene Aliases: Gprk6; Grk6
          View more View less
          UniProt ID: (Mouse) O70293
          View more View less
          Entrez Gene ID: (Mouse) 26385
          View more View less
          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity G-protein coupled receptor kinase activity protein binding ATP binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups beta-adrenergic receptor kinase activity non-receptor serine/threonine protein kinase
          Process(es)
          desensitization of G-protein coupled receptor protein signaling pathway protein phosphorylation signal transduction negative regulation of anion channel activity Wnt signaling pathway phosphorylation termination of G-protein coupled receptor signaling pathway
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline