          • Primary Antibodies ›
          • PCDHGA3 Antibodies
          Antibodies Inc
          Gamma-Protocadherin-A3 Monoclonal Antibody (N144/17)
          View all (6) PCDHGA3 antibodies
          Tech Support
          Cite Gamma-Protocadherin-A3 Monoclonal Antibody (N144/17)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Gamma-Protocadherin-A3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Gamma-Protocadherin-A3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Gamma-Protocadherin-A3 Antibody (75-177) in WB
          Transfected cell immunoblot: extracts of COS cells transiently transfected with GFP-tagged Gamma- protocadherin-A3 and -A4 and untagged Kv2.1 plasmids and probed with N144/17 (left) and N159/5 (right) TC supe. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Gamma-Protocadherin-A3 Antibody in Western Blot (WB)
          Product Details
          75-177
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N144/17
          Immunogen
          Fusion protein amino acids 720-804 (variable cytoplasmic domain) of mouse Gamma-protocadherin-A3 (accession number Q91XY5) produced recombinantly in E. Coli.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
          Bioinformatics
          Protein Aliases: Gamma-Protocadherin-A3; PCDG3; PCDH gamma A3; protocadherin gamma-A3
          View more View less
          Entrez Gene ID: (Mouse) 93711
          View more View less
          Function(s)
          protein binding cadherin
          Process(es)
          endosome to lysosome transport lipid tube assembly involved in organelle fusion
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0