    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • LRP4 Antibodies
          Antibodies Inc
          LRP4 (Cytoplasmic) Monoclonal Antibody (N164/6), FL594
          View all (28) LRP4 antibodies
          Tech Support
          Cite LRP4 (Cytoplasmic) Monoclonal Antibody (N164/6), FL594
          Additional Information:
          {{banner.description}}
          LRP4 (Cytoplasmic) Monoclonal Antibody (N164/6), FL594
          Product Details
          75-182-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N164/6
          Immunogen
          Fusion protein amino acids 1747-1905 (cytoplasmic C-terminus) of mouse LRP4 (accession number Q8VI56) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          This gene encodes a member of the low-density lipoprotein receptor-related protein family. The encoded protein may be a regulator of Wnt signaling. Mutations in this gene are associated with Cenani-Lenz syndrome.
          Bioinformatics
          Protein Aliases: Corin; LDLR; LDLR dan; Low-density lipoprotein receptor-related protein 4; LRP-4; multiple epidermal growth factor-like domains 7
          View more View less
          Gene Aliases: 6430526J12Rik; D230026E03; Kiaa0816; Lrp4; mdig; Megf7
          View more View less
          UniProt ID: (Mouse) Q8VI56
          View more View less
          Entrez Gene ID: (Rat) 83469, (Mouse) 228357
          View more View less
          Function(s)
          calcium ion binding receptor tyrosine kinase binding apolipoprotein binding protein homodimerization activity scaffold protein binding protein binding Wnt-protein binding Wnt-activated receptor activity apolipoprotein
          Process(es)
          kidney development regulation of protein phosphorylation hair follicle development endocytosis protein localization dorsal/ventral pattern formation proximal/distal pattern formation Wnt signaling pathway negative regulation of Wnt signaling pathway negative regulation of ossification embryonic limb morphogenesis odontogenesis of dentin-containing tooth embryonic digit morphogenesis receptor clustering dendrite morphogenesis anatomical structure development negative regulation of axonogenesis synapse organization synaptic growth at neuromuscular junction protein heterotetramerization limb development skeletal muscle acetylcholine-gated channel clustering negative regulation of canonical Wnt signaling pathway postsynaptic membrane assembly presynaptic membrane assembly positive regulation of presynaptic membrane organization multicellular organism development cell differentiation Wnt signaling pathway involved in dorsal/ventral axis specification canonical Wnt signaling pathway
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline