    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • QKI-7 Antibodies
          Antibodies Inc
          QKI-7 Monoclonal Antibody (N183/15)
          View all (6) QKI-7 antibodies
          Tech Support
          Cite QKI-7 Monoclonal Antibody (N183/15)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          QKI-7 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          QKI-7 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          QKI-7 Antibody (75-200) in IHC (F)
          Double immunofluorescence revealing pruned Lewy Neurites (pSyn, Red) and the cytoplasms of mature mouse OLs (QKI-7, cat. 75-200, 1:500; green) at the level of the anterior commissure of a mouse 6 months after the intrastriatal injection of alpha--Syn PFFs. Horizontal brain section. Nuclei are counterstained with DRAQ7 (white). Image from publication CC-BY-4.0. PMID:36830639. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          QKI-7 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          QKI-7 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          QKI-7 Antibody in Western Blot (WB)
          QKI-7 Antibody in Western Blot (WB)
          QKI-7 Antibody in Western Blot (WB)
          QKI-7 Monoclonal Antibody (N183/15)
          Product Details
          75-200
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          1:200
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N183/15
          Immunogen
          Synthetic peptide amino acids 303-317 of human QKI-7 (accession number Q96PU8-5).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: LEWIEMPVMP DISAH
          Target Information
          QKI-7 is an isoform of the Quaking (QKI) protein, which belongs to the STAR (Signal Transduction and Activation of RNA) family of RNA-binding proteins. QKI proteins play essential roles in RNA metabolism, including RNA splicing, stabilization, and translation, and are crucial for various cellular processes. QKI-7 is an important RNA-binding protein with significant roles in the nervous system, particularly in the regulation of myelination and neuronal RNA processing. Its study is essential for understanding the molecular basis of various neurological disorders.
          Bioinformatics
          Protein Aliases: homolog of mouse quaking QKI (KH domain RNA binding protein); Hqk; KH domain-containing RNA-binding protein QKI; lethal, Chr 17, U Wisconsin 1; MqkI; Protein quaking; QKI-5 protein; QKI/LOC100132735 fusion; quaking; quaking homolog, KH domain RNA binding; QUAKING-3; RNA binding protein HQK; unnamed protein product
          View more View less
          Gene Aliases: 1110003F05Rik; 1500005P18; HKQ; Hqk; hqkI; l(17)-1Wis; l17Wis1; QK; QK1; QK3; QKI
          View more View less
          UniProt ID: (Human) Q96PU8, (Mouse) Q9QYS9
          View more View less
          Entrez Gene ID: (Human) 9444, (Mouse) 19317
          View more View less
          Function(s)
          nucleic acid binding DNA binding transcription coactivator activity RNA binding mRNA binding mRNA 3'-UTR binding protein binding SH3 domain binding miRNA binding internal N(7)-methylguanine-containing RNA reader activity poly(A) RNA binding RNA splicing factor
          Process(es)
          vasculogenesis mRNA processing regulation of translation spermatid development intracellular mRNA localization axon ensheathment RNA splicing positive regulation of gene expression regulation of epithelial to mesenchymal transition microglia differentiation negative regulation of angiogenesis negative regulation of translation cell differentiation positive regulation of myelination negative regulation of type I interferon production stress granule assembly vascular smooth muscle cell differentiation myelination long-chain fatty acid biosynthetic process positive regulation of cholesterol biosynthetic process regulation of macrophage differentiation negative regulation of macrophage differentiation positive regulation of transcription, DNA-templated regulation of mRNA splicing, via spliceosome mRNA stabilization regulation of astrocyte differentiation positive regulation of oligodendrocyte differentiation mRNA transport regulation of cell development negative regulation of cold-induced thermogenesis spliceosome-depend formation of circular RNA negative regulation of 3'-UTR-mediated mRNA stabilization myofibroblast contraction regulation of multicellular organismal development negative regulation of miRNA catabolic process transport multicellular organism development muscle cell differentiation 3'-UTR-mediated mRNA destabilization
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline