Sequence of this protein is as follows: LEWIEMPVMP DISAH
QKI-7 is an isoform of the Quaking (QKI) protein, which belongs to the STAR (Signal Transduction and Activation of RNA) family of RNA-binding proteins. QKI proteins play essential roles in RNA metabolism, including RNA splicing, stabilization, and translation, and are crucial for various cellular processes. QKI-7 is an important RNA-binding protein with significant roles in the nervous system, particularly in the regulation of myelination and neuronal RNA processing. Its study is essential for understanding the molecular basis of various neurological disorders.
Protein Aliases: homolog of mouse quaking QKI (KH domain RNA binding protein); Hqk; KH domain-containing RNA-binding protein QKI; lethal, Chr 17, U Wisconsin 1; MqkI; Protein quaking; QKI-5 protein; QKI/LOC100132735 fusion; quaking; quaking homolog, KH domain RNA binding; QUAKING-3; RNA binding protein HQK; unnamed protein product
Gene Aliases: 1110003F05Rik; 1500005P18; HKQ; Hqk; hqkI; l(17)-1Wis; l17Wis1; QK; QK1; QK3; QKI
UniProt ID: (Human) Q96PU8, (Mouse) Q9QYS9
Entrez Gene ID: (Human) 9444, (Mouse) 19317