    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • RNF146 Antibodies
          Antibodies Inc
          Iduna/RNF146 Monoclonal Antibody (N201/35), FL650
          View all (13) RNF146 antibodies
          Questions & Answers
          Cite Iduna/RNF146 Monoclonal Antibody (N201/35), FL650
          Iduna/RNF146 Monoclonal Antibody (N201/35), FL650
          Product Details
          75-233-FL650
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N201/35
          Immunogen
          Fusion protein amino acids 1-359 (full-length) of mouse Iduna (accession number Q9CZW6) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          RNF146 protein function is currently unknown.
          Bioinformatics
          Protein Aliases: E3 ubiquitin-protein ligase RNF146; Iduna; RING finger protein 146; RING-type E3 ubiquitin transferase RNF146
          View more View less
          Gene Aliases: 2610509H23Rik; Iduna; Rnf146
          View more View less
          UniProt ID: (Rat) Q5XIK5, (Mouse) Q3TF93
          View more View less
          Entrez Gene ID: (Rat) 308051, (Mouse) 68031
          View more View less
          Function(s)
          ubiquitin-protein transferase activity zinc ion binding ligase activity poly-ADP-D-ribose binding UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity ribosomal S6-glutamic acid ligase activity coenzyme F420-0 gamma-glutamyl ligase activity coenzyme F420-2 alpha-glutamyl ligase activity metal ion binding protein-glycine ligase activity protein-glycine ligase activity, initiating protein-glycine ligase activity, elongating tubulin-glycine ligase activity protein-glutamic acid ligase activity tubulin-glutamic acid ligase activity
          Process(es)
          Wnt signaling pathway protein ubiquitination involved in ubiquitin-dependent protein catabolic process protein autoubiquitination protein K48-linked ubiquitination positive regulation of canonical Wnt signaling pathway
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline