    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PEX5L Antibodies
          Antibodies Inc
          TRIP8b (Exon 1b) Monoclonal Antibody (N212A/34)
          View all (30) PEX5L antibodies
          Tech Support
          Cite TRIP8b (Exon 1b) Monoclonal Antibody (N212A/34)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          TRIP8b (Exon 1b) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TRIP8b (Exon 1b) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          TRIP8b (Exon 1b) Antibody (75-245) in WB
          Transfected cell immunoblot: extracts of COS cells transiently transfected with GFP-tagged TRIP8b-1a-4, TRIP8b-1b-2, TRIP8b-1a-2 or untagged Kv2.1 plasmid and probed with N212A/34 (left) or N86/8 (right) TC supe. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRIP8b (Exon 1b) Antibody in Western Blot (WB)
          TRIP8b (Exon 1b) Antibody in Western Blot (WB)
          TRIP8b (Exon 1b) Antibody in Western Blot (WB)
          TRIP8b (Exon 1b) Monoclonal Antibody (N212A/34)
          Product Details
          75-245
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N212A/34
          Immunogen
          Fusion protein amino acids 1-192 (exon 1a, exon 4 and constant region) of rat TRIP8b (accession number Q925N3) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Pex5l is an accessory subunit of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels, regulating their cell-surface expression and cyclic nucleotide dependence. Research studies have shown that Pex5l is expressed in the brain, testis and pancreas. Diseases associated with PEX5L include Rhizomelic Chondrodysplasia Punctata, Type 5 and Rhizomelic Chondrodysplasia Punctata, Type 2.
          Bioinformatics
          Protein Aliases: HCN channel auxiliary subunit; peroxin 2; Peroxin-5-related protein; peroxisomal targeting signal 1 receptor-like; Peroxisome biogenesis factor 5-like; PEX2-related protein; PEX5-like protein; PEX5-related protein; Pex5p-related protein; Pex5Rp; Tetratricopeptide repeat-containing Rab8b-interacting protein; TPR-containing Rab8b-interacting protein; unnamed protein product
          View more View less
          Gene Aliases: 1700016J08Rik; Pex2; PEX5L; PEX5R; PEX5RP; PXR2; PXR2B; TRIP8b; Trjip8b
          View more View less
          UniProt ID: (Human) Q8IYB4, (Rat) Q925N3, (Mouse) Q8C437
          View more View less
          Entrez Gene ID: (Human) 51555, (Rat) 286937, (Mouse) 58869
          View more View less
          Function(s)
          peroxisome targeting sequence binding peroxisome matrix targeting signal-1 binding protein binding small GTPase binding intracellular cyclic nucleotide activated cation channel activity Rab GTPase binding membrane trafficking regulatory protein
          Process(es)
          protein import into peroxisome matrix, docking cation transport protein import into peroxisome matrix regulation of membrane potential regulation of cAMP-mediated signaling regulated exocytosis maintenance of protein location positive regulation of corticotropin secretion cation transmembrane transport
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline