    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PEX5L Antibodies
          Antibodies Inc
          TRIP8b (Exon 1b) Monoclonal Antibody (N212A/34), FL594
          View all (31) PEX5L antibodies
          Tech Support
          Cite TRIP8b (Exon 1b) Monoclonal Antibody (N212A/34), FL594
          Additional Information:
          {{banner.description}}
          TRIP8b (Exon 1b) Monoclonal Antibody (N212A/34), FL594
          Product Details
          75-245-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N212A/34
          Immunogen
          Fusion protein amino acids 1-192 (exon 1a, exon 4 and constant region) of rat TRIP8b (accession number Q925N3) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Pex5l is an accessory subunit of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels, regulating their cell-surface expression and cyclic nucleotide dependence. Research studies have shown that Pex5l is expressed in the brain, testis and pancreas. Diseases associated with PEX5L include Rhizomelic Chondrodysplasia Punctata, Type 5 and Rhizomelic Chondrodysplasia Punctata, Type 2.
          Bioinformatics
          Protein Aliases: HCN channel auxiliary subunit; peroxin 2; Peroxin-5-related protein; peroxisomal targeting signal 1 receptor-like; Peroxisome biogenesis factor 5-like; PEX2-related protein; PEX5-like protein; PEX5-related protein; Pex5p-related protein; Pex5Rp; Tetratricopeptide repeat-containing Rab8b-interacting protein; TPR-containing Rab8b-interacting protein; unnamed protein product
          View more View less
          Gene Aliases: 1700016J08Rik; Pex2; PEX5L; PEX5R; PEX5RP; PXR2; PXR2B; TRIP8b; Trjip8b
          View more View less
          UniProt ID: (Human) Q8IYB4, (Rat) Q925N3, (Mouse) Q8C437
          View more View less
          Entrez Gene ID: (Human) 51555, (Rat) 286937, (Mouse) 58869
          View more View less
          Function(s)
          peroxisome targeting sequence binding peroxisome matrix targeting signal-1 binding protein binding small GTPase binding intracellular cyclic nucleotide activated cation channel activity Rab GTPase binding membrane trafficking regulatory protein
          Process(es)
          protein import into peroxisome matrix, docking cation transport protein import into peroxisome matrix regulation of membrane potential regulation of cAMP-mediated signaling regulated exocytosis maintenance of protein location positive regulation of corticotropin secretion cation transmembrane transport
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline