    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FGF13B Isoform Antibodies
          Antibodies Inc
          FGF13/FHF2/B Isoform Monoclonal Antibody (N225A/10), FL490
          View all (5) FGF13B Isoform antibodies
          Tech Support
          Cite FGF13/FHF2/B Isoform Monoclonal Antibody (N225A/10), FL490
          Additional Information:
          {{banner.description}}
          FGF13/FHF2/B Isoform Monoclonal Antibody (N225A/10), FL490
          Product Details
          75-248-FL490
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N225A/10
          Immunogen
          Synthetic peptide amino acids 1-20 of human FHF2B (accession number Q92913-2).
          View immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: MALLRKSYSE PQLKGIVTKL
          Target Information
          FGF13B is an isoform of the fibroblast growth factor 13 (FGF13) protein, which belongs to the intracellular FGF family. This isoform plays a crucial role in the regulation of ion channels, neuronal development, and intracellular signaling pathways. Dysregulation of FGF13B has been implicated in various neurological conditions, including intellectual disabilities and epilepsy.
          Bioinformatics
          Protein Aliases: FGF-13; FHF-2; FHF2-1A; FHF2-1B; Fibroblast growth factor 13; Fibroblast growth factor homologous factor 2; hFHF-2(1X+1W+1V); hFHF-2(1Z+1Y); unnamed protein product
          View more View less
          Gene Aliases: DEE90; FGF-13; FGF13; FGF2; FHF-2; FHF2; LINC00889; XLID110
          View more View less
          UniProt ID: (Human) Q92913
          View more View less
          Entrez Gene ID: (Human) 2258
          View more View less
          Function(s)
          protein binding microtubule binding growth factor activity sodium channel regulator activity ion channel binding beta-tubulin binding
          Process(es)
          MAPK cascade neuron migration sodium ion transport negative regulation of microtubule depolymerization signal transduction cell-cell signaling nervous system development learning or memory learning memory hippocampus development cerebral cortex cell migration neurogenesis establishment of neuroblast polarity microtubule polymerization negative regulation of collateral sprouting branching morphogenesis of a nerve protein localization to plasma membrane regulation of cardiac muscle cell action potential involved in regulation of contraction inhibitory synapse assembly positive regulation of voltage-gated sodium channel activity
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0