    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • FGF13B Isoform Antibodies
          Antibodies Inc
          FGF13/FHF2/B Isoform Monoclonal Antibody (N225A/10), FL594
          View all (5) FGF13B Isoform antibodies
          Tech Support
          Cite FGF13/FHF2/B Isoform Monoclonal Antibody (N225A/10), FL594
          Additional Information:
          {{banner.description}}
          FGF13/FHF2/B Isoform Monoclonal Antibody (N225A/10), FL594
          Product Details
          75-248-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N225A/10
          Immunogen
          Synthetic peptide amino acids 1-20 of human FHF2B (accession number Q92913-2).
          View immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: MALLRKSYSE PQLKGIVTKL
          Target Information
          FGF13B is an isoform of the fibroblast growth factor 13 (FGF13) protein, which belongs to the intracellular FGF family. This isoform plays a crucial role in the regulation of ion channels, neuronal development, and intracellular signaling pathways. Dysregulation of FGF13B has been implicated in various neurological conditions, including intellectual disabilities and epilepsy.
          Bioinformatics
          Protein Aliases: FGF-13; FHF-2; FHF2-1A; FHF2-1B; Fibroblast growth factor 13; Fibroblast growth factor homologous factor 2; hFHF-2(1X+1W+1V); hFHF-2(1Z+1Y); unnamed protein product
          View more View less
          Gene Aliases: DEE90; FGF-13; FGF13; FGF2; FHF-2; FHF2; LINC00889; XLID110
          View more View less
          UniProt ID: (Human) Q92913
          View more View less
          Entrez Gene ID: (Human) 2258
          View more View less
          Function(s)
          protein binding microtubule binding growth factor activity sodium channel regulator activity ion channel binding beta-tubulin binding
          Process(es)
          MAPK cascade neuron migration sodium ion transport negative regulation of microtubule depolymerization signal transduction cell-cell signaling nervous system development learning or memory learning memory hippocampus development cerebral cortex cell migration neurogenesis establishment of neuroblast polarity microtubule polymerization negative regulation of collateral sprouting branching morphogenesis of a nerve protein localization to plasma membrane regulation of cardiac muscle cell action potential involved in regulation of contraction inhibitory synapse assembly positive regulation of voltage-gated sodium channel activity
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline