    • 엔터프라이즈급 실험실 정보 처리
    • 360° CDMO 및 CRO 서비스
    • CDMO 서비스
    • 임상시험 CRO 서비스
    • 장비 서비스
    • 교육 서비스
    • Unity Lab Services
    • 서비스 전체 보기
  • 지원
    • 고객센터
    • 문의하기
    • 시험성적서(COA) 및 적합성 인증서(COC)
    • Safety Data Sheets (SDS)
    • 매뉴얼
    • 인용 및 참고 문헌
    • 기기 지원
    • 기술 자료/제품 FAQ
    • 학습 센터
    • 지원 메뉴 전체보기
      Cell Culture Plastics
  • 문의하기
  • 빠른 주문
  • 주문 현황
  • 제품 문서 검색
Thermo Fisher Scientific Logo
Search
전체검색
Search button
          • 주문 현황
          • 빠른주문
          • 로그인
            로그인
            회원이 아니신가요? 계정 생성하기
            • 계정정보
            • 주문내역조회
            • 커스텀제품 및 프로젝트
            • Services Central
          • Primary Antibodies ›
          • Brevican Antibodies
          Antibodies Inc
          Brevican Monoclonal Antibody (N294A/6)
          View all (23) Brevican antibodies
          Tech Support
          Cite Brevican Monoclonal Antibody (N294A/6)
          Additional Information:
          {{banner.description}}
          Brevican Monoclonal Antibody (N294A/6)
          Product Details
          75-281
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          Assay-dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N294A/6
          Immunogen
          Fusion protein amino acids 219-655 of rat Brevican (accession number P55068) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • FL490
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4
          Contains
          0.065% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Brevican is the most abundant chondroitin sulfate proteoglycan in the extracellular matrix of the adult brain. It is a member of the lectican family of aggregating extracellular matrix proteoglycans that bear chondroitin sulfate (CS) chains. It is highly expressed in the central nervous system and is thought to stabilize synapses and inhibit neural plasticity. Brevican is secreted from astrocytes and neurons as a 145 kD core protein that bears up to three, covalently-linked, CS chains. It is also is secreted as a 145 kD core protein without CS chains. When cleaved by extracellular glutamyl endopeptidases, the ADAMTSs, a 55 kD N-terminal fragment is formed that contains the unique C terminal epitope EAMESE.
          Bioinformatics
          Protein Aliases: BEHAB; Brain-enriched hyaluronan-binding protein; brevican (brain specific proteoglycan in the aggrecan family); Brevican core protein
          View more View less
          Gene Aliases: ALPBRE; Bcan; BEHAB; Cspg7
          View more View less
          UniProt ID: (Rat) P55068, (Mouse) Q61361
          View more View less
          Entrez Gene ID: (Rat) 25393, (Mouse) 12032
          View more View less
          Function(s)
          extracellular matrix structural constituent hyaluronic acid binding carbohydrate binding extracellular matrix glycoprotein
          Process(es)
          skeletal system development cell adhesion central nervous system development hippocampus development regulation of neuronal synaptic plasticity
          Contact tech support
          온라인 주문 Plus Icon Minus Icon
          • 주문 현황
          • 주문 지원
          • 빠른 주문
          • Supply Center
          • eProcurement
          지원 Plus Icon Minus Icon
          • Help and Support
          • 고객 센터
          • 기술 지원 센터
          • 제품 문서 검색
          • 사이트 문제 보고
          교육 및 이벤트 Plus Icon Minus Icon
          • 교육 센터
          • 프로모션
          • 이벤트 및 웨비나
          • 소셜 미디어
          • 소개 소개
          • 채용 채용
          • 투자자 투자자
          • 뉴스 뉴스
          • 사회적 책임 사회적 책임
          • Trademarks
          • 공정거래 공정거래
          • 정책 및 고지
          • PPD
          • 이용 약관
          • 개인정보 처리방침
          • 가격 및 운임 정책
          Korea flag icon
          Korea
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          고객센터 문의 | 평일 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          장비 서비스 문의 | 평일 09:00~18:00
          1661-5055   |   Live Chat
          써모 피셔 사이언티픽 코리아 주식회사
          대표자 : 석수진
          사업자 등록번호 : 117-81-46910
          입금계좌 : 하나은행 336-890014-06204
          (예금주 : 써모피셔사이언티픽코리아 주식회사)
          써모 피셔 사이언티픽 솔루션스 유한회사
          대표자 : 석수진
          사업자 등록번호 : 114-86-04783
          입금계좌 : 신한은행 140-004-396660
          (예금주 : 써모피셔사이언티픽솔루션스 유한회사)
          주소 : 서울시 강남구 광평로 281 수서오피스빌딩 12층 06349 | 통신판매업신고번호 : 2015-서울강남-00898
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0